This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01387734_cn
See other MTHFR CNV Assays ›
Gene Symbol
MTHFR
Assay Reference Genome
Location

Chr.1:11801251 on build GRCh38
Cytoband
1p36.22
Assay ID Hs01387734_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

4524

Gene Name:

methylenetetrahydrofolate reductase (NAD(P)H)

Gene Aliases:

-

Location:

Chr.1:11785730-11806103 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MTHFR NM_005957.4 3 614 NP_005948.3
XM_005263458.3 3 1451 XP_005263515.1
XM_005263460.4 3 1069 XP_005263517.1
XM_005263462.4 3 502 XP_005263519.1
XM_005263463.3 3 631 XP_005263520.1
XM_011541495.2 3 1448 XP_011539797.1
XM_011541496.2 3 1451 XP_011539798.1
XM_017001328.1 3 1451 XP_016856817.1
AB209113.1 2 652 BAD92350.1
AJ237672.1 2 514 CAB41971.1
AK312907.1 3 475
BC053509.1 3 436 AAH53509.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv518652 Chr.1:11797654 - 11802721 on Build GRCh38 Loss MTHFR

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cellular amino acid metabolic process
methionine metabolic process
blood circulation
regulation of histone methylation
response to vitamin B2
tetrahydrofolate interconversion
response to drug
response to amino acid
S-adenosylmethionine metabolic process
folic acid metabolic process
homocysteine metabolic process
response to folic acid
oxidation-reduction process
response to interleukin-1
heterochromatin maintenance
methylenetetrahydrofolate reductase (NAD(P)H) activity
protein complex binding
flavin adenine dinucleotide binding
NADP binding
modified amino acid binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline