This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01439443_cn
See other FAM118B CNV Assays ›
Gene Symbol
FAM118B
Assay Reference Genome
Location

Chr.11:126211541 on build GRCh38
Cytoband
11q24.2
Assay ID Hs01439443_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79607

Gene Name:

family with sequence similarity 118 member B

Gene Aliases:

-

Location:

Chr.11:126211414-126264540 on Build GRCh38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FAM118B XM_005271665.3 1 128 XP_005271722.1
XM_011542978.2 1 128 XP_011541280.1
XM_017018286.1 1 128 XP_016873775.1

Target Gene Details

Entrez Gene ID:

84881

Gene Name:

RNA pseudouridylate synthase domain containing 4

Gene Aliases:

-

Location:

Chr.11:126202094-126211692 on Build GRCh38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
RPUSD4 NM_001144827.1 1 152 NP_001138299.1
NM_032795.2 1 152 NP_116184.2
XM_011543039.2 1 122 XP_011541341.1
AK027400.1 1 106 BAB55086.1
AK225320.1 1 112
AK308059.1 1 106
BC014131.2 1 113 AAH14131.1
DB051034.1 1 152

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv556529 Chr.11:126204297 - 126264724 on Build GRCh38 Loss FAM118B RPUSD4 SRPRA
nsv556527 Chr.11:126172311 - 126211508 on Build GRCh38 Loss FAM118B RPUSD4

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

Cajal body organization
tRNA pseudouridine synthesis
protein binding
pseudouridine synthase activity
deaminase activity
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline