This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01467476_cn
See other PPP1R16A CNV Assays ›
Gene Symbol
PPP1R16A
Assay Reference Genome
Location

Chr.8:144497458 on build GRCh38
Cytoband
8q24.3
Assay ID Hs01467476_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

84988

Gene Name:

protein phosphatase 1 regulatory subunit 16A

Gene Aliases:

MYPT3

Location:

Chr.8:144477987-144502121 on Build GRCh38

Assay Gene Location:

Overlaps Exon 5 - Intron 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
PPP1R16A XM_005272350.2 XP_005272407.1
XM_011517349.2 XP_011515651.1
XM_017013912.1 XP_016869401.1
XM_017013913.1 XP_016869402.1
XM_017013914.1 XP_016869403.1
XM_017013915.1 XP_016869404.1
AK090471.1 4 1132 BAC03452.1
BC007854.2 AAH07854.1
BC049841.1 AAH49841.1
BC053506.1 AAH53506.1
BT006845.1 AAP35491.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv470249 Chr.8:143889098 - 144519234 on Build GRCh38 Loss PARP10 SPATC1 ADCK5 KIFC2 HSF1 SCX MIR661 MIR6847 MIR1234 CPSF1 GPT CYC1 MIR6848 LOC101928902 SHARPIN FOXH1 DGAT1 LRRC14 SCRT1 GPAA1 OPLAH MROH1 BOP1 PPP1R16A MAF1 SLC39A4 TONSL-AS1 LOC101928953 MIR6893 VPS28 PLEC EXOSC4 GRINA HGH1 CYHR1 RECQL4 SLC52A2 SMPD5 MIR6849 TONSL MFSD3 TMEM249 FBXL6 MIR939 MIR6846 MIR7112 WDR97
nsv612951 Chr.8:144479830 - 144677692 on Build GRCh38 Loss LRRC14 RECQL4 LOC101928953 LRRC24 C8orf82 MFSD3 PPP1R16A GPT ARHGAP39
nsv831489 Chr.8:144342140 - 144544026 on Build GRCh38 Loss ADCK5 KIFC2 MIR1234 CPSF1 GPT LOC101928902 FOXH1 LRRC14 LRRC24 PPP1R16A SLC39A4 TONSL-AS1 LOC101928953 C8orf82 MIR6893 VPS28 ARHGAP39 CYHR1 RECQL4 SLC52A2 MIR6849 TONSL MFSD3 TMEM249 FBXL6 MIR939
dgv12439n54 Chr.8:144438306 - 144525122 on Build GRCh38 Loss FOXH1 CYHR1 TONSL-AS1 LRRC14 KIFC2 RECQL4 LOC101928953 LRRC24 TONSL MFSD3 PPP1R16A GPT
nsv612955 Chr.8:144494912 - 144502322 on Build GRCh38 Loss LOC101928953 PPP1R16A GPT
nsv482126 Chr.8:144496726 - 144502120 on Build GRCh38 Gain LOC101928953 PPP1R16A
dgv907n27 Chr.8:144416180 - 144525122 on Build GRCh38 Loss TONSL-AS1 KIFC2 LOC101928953 MIR6893 VPS28 GPT FOXH1 CYHR1 LRRC14 RECQL4 LRRC24 TONSL MFSD3 PPP1R16A SLC39A4
dgv12442n54 Chr.8:144492696 - 144516381 on Build GRCh38 Loss RECQL4 LOC101928953 MFSD3 PPP1R16A GPT
dgv12434n54 Chr.8:144270080 - 144516801 on Build GRCh38 Loss ADCK5 KIFC2 HSF1 MIR1234 CPSF1 GPT MIR6848 LOC101928902 FOXH1 DGAT1 SCRT1 BOP1 PPP1R16A SLC39A4 TONSL-AS1 LOC101928953 MIR6893 VPS28 CYHR1 RECQL4 SLC52A2 MIR6849 TONSL MFSD3 TMEM249 FBXL6 MIR939
nsv515809 Chr.8:143904439 - 144519234 on Build GRCh38 Gain+Loss PARP10 SPATC1 ADCK5 KIFC2 HSF1 SCX MIR661 MIR6847 MIR1234 CPSF1 GPT CYC1 MIR6848 LOC101928902 SHARPIN FOXH1 DGAT1 LRRC14 SCRT1 GPAA1 OPLAH MROH1 BOP1 PPP1R16A MAF1 SLC39A4 TONSL-AS1 LOC101928953 MIR6893 VPS28 PLEC EXOSC4 GRINA HGH1 CYHR1 RECQL4 SLC52A2 SMPD5 MIR6849 TONSL MFSD3 TMEM249 FBXL6 MIR939 MIR6846 MIR7112 WDR97
nsv612940 Chr.8:144436292 - 144613673 on Build GRCh38 Loss TONSL-AS1 KIFC2 LOC101928953 C8orf82 GPT ARHGAP39 FOXH1 CYHR1 LRRC14 RECQL4 LRRC24 TONSL MFSD3 PPP1R16A
nsv428208 Chr.8:144041826 - 144722291 on Build GRCh38 Gain SPATC1 ADCK5 KIFC2 HSF1 SCX MIR6847 MIR1234 CPSF1 GPT CYC1 MIR6848 LOC101928902 SHARPIN FOXH1 DGAT1 LRRC14 LRRC24 SCRT1 GPAA1 OPLAH MROH1 BOP1 PPP1R16A MAF1 SLC39A4 TONSL-AS1 LOC101928953 C8orf82 MIR6893 VPS28 EXOSC4 ZNF251 ARHGAP39 HGH1 CYHR1 RECQL4 SLC52A2 SMPD5 MIR6849 TONSL MFSD3 TMEM249 FBXL6 MIR939 MIR6846 MIR7112 WDR97
esv29999 Chr.8:144456732 - 144525122 on Build GRCh38 Loss FOXH1 CYHR1 LRRC14 KIFC2 RECQL4 LOC101928953 LRRC24 MFSD3 PPP1R16A GPT

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

phosphatase modulator

Gene Ontology Categories:

Function(s) Process(es)

regulation of catalytic activity
protein binding
protein phosphatase type 1 regulator activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-ndfxr:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline