This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01480084_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1257315 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01480084_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Overlaps Intron 6 - Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss MIR6726 ACAP3 UBE2J2 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication CPSF3L MIR6726 TNFRSF4 TTLL10 TTLL10-AS1 RNF223 C1orf159 LOC100288175 UBE2J2 LINC01342 MIR429 TNFRSF18 AGRN ACAP3 MIR6727 LOC105378948 SCNN1D CPTP B3GALT6 MIR200A MIR200B FAM132A SDF4 PUSL1 TAS1R3
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss NOC2L CPSF3L LOC100287934 MIR6726 LOC107985729 LINC00115 LOC148413 DVL1 MMP23B TTLL10 TTLL10-AS1 SLC35E2 SLC35E2B LOC284600 TMEM240 AURKAIP1 UBE2J2 CDK11B LINC01342 FNDC10 SSU72 ATAD3A ATAD3C ACAP3 MIR6727 MXRA8 CCNL2 FAM41C LOC102724312 MRPL20 MMP23A CPTP FAM87B TAS1R3 SAMD11 VWA1 LOC100288069 ATAD3B TNFRSF4 RNF223 C1orf159 PERM1 KLHL17 NADK LOC100288175 CDK11A ISG15 ANKRD65 MIB2 MIR429 TNFRSF18 AGRN MIR6808 LOC105378948 SCNN1D B3GALT6 LINC01128 MIR200A PLEKHN1 LOC107985728 MIR200B FAM132A SDF4 PUSL1 TMEM88B LOC100130417 HES4
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CPSF3L LOC100287934 MIR6723 OR4F5 LOC148413 TTLL10 SLC35E2 SLC35E2B LOC284600 UBE2J2 LOC100506504 CDK11B FNDC10 ATAD3A MORN1 ACAP3 MIR6727 MXRA8 LOC105378949 FAM41C MRPL20 MTND1P23 MMP23A DDX11L1 VWA1 LOC100288069 TNFRSF4 RNF223 C1orf159 SKI KLHL17 OR4F16 ISG15 GABRD ANKRD65 MTND2P28 LOC105378592 WASH7P TMEM52 LOC105378589 AGRN LOC729737 LOC100133331 CALML6 B3GALT6 LINC01128 MIR200B FAM132A SDF4 PUSL1 LOC105378591 NOC2L MIR6726 LOC100129534 LOC107985729 OR4F29 LINC00115 DVL1 MMP23B TTLL10-AS1 LOC102725121 TMEM240 AURKAIP1 GNB1 MIR1302-2 LINC01342 SSU72 MIR6859-1 ATAD3C CCNL2 LOC102724312 CPTP LOC100134822 FAM87B TAS1R3 SAMD11 ATAD3B PRKCZ MIR6859-2 PERM1 FAAP20 NADK LOC100288175 CDK11A LOC107984841 LOC105378947 LOC100132287 MIB2 MIR429 TNFRSF18 LOC101928626 MIR6808 LOC105378948 SCNN1D MIR200A CFAP74 PLEKHN1 LOC107985728 FAM138A TMEM88B LOC100130417 HES4
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss MIR6726 ACAP3 FAM132A SDF4 UBE2J2 SCNN1D B3GALT6
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss NOC2L TNFRSF4 LINC00115 TTLL10 TTLL10-AS1 RNF223 C1orf159 PERM1 KLHL17 LOC100288175 LOC284600 ISG15 UBE2J2 LINC01342 MIR429 TNFRSF18 AGRN FAM41C LOC105378948 B3GALT6 LINC01128 MIR200A PLEKHN1 LOC107985728 MIR200B FAM132A FAM87B SDF4 LOC100130417 HES4 SAMD11
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion VWA1 CPSF3L MIR6726 ATAD3B LOC107985729 TNFRSF4 LOC148413 DVL1 MMP23B TTLL10 TTLL10-AS1 C1orf159 TMEM240 AURKAIP1 UBE2J2 CDK11B LINC01342 ANKRD65 FNDC10 MIB2 MIR429 TNFRSF18 SSU72 ATAD3A ATAD3C ACAP3 MIR6727 MXRA8 CCNL2 MIR6808 LOC102724312 MRPL20 SCNN1D CPTP B3GALT6 MIR200A MIR200B FAM132A SDF4 PUSL1 TAS1R3 TMEM88B
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain NOC2L CPSF3L MIR6726 TNFRSF4 LOC148413 DVL1 TTLL10 TTLL10-AS1 RNF223 C1orf159 PERM1 KLHL17 LOC100288175 LOC284600 AURKAIP1 ISG15 UBE2J2 LINC01342 ANKRD65 MIR429 TNFRSF18 AGRN ACAP3 MIR6727 MXRA8 CCNL2 MIR6808 MRPL20 LOC105378948 SCNN1D CPTP B3GALT6 MIR200A PLEKHN1 LOC107985728 MIR200B FAM132A SDF4 PUSL1 TAS1R3 TMEM88B LOC100130417 HES4 SAMD11
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain MIR429 TNFRSF18 TNFRSF4 TTLL10 TTLL10-AS1 LOC105378948 SCNN1D RNF223 C1orf159 B3GALT6 MIR200A MIR200B FAM132A SDF4 UBE2J2 LINC01342
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR6726 MIR429 TNFRSF18 TNFRSF4 ACAP3 TTLL10 TTLL10-AS1 SCNN1D RNF223 C1orf159 B3GALT6 MIR200A MIR200B FAM132A SDF4 UBE2J2 LINC01342
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain CPSF3L MIR6726 MIR429 TNFRSF18 TNFRSF4 ACAP3 MIR6727 TTLL10 TTLL10-AS1 SCNN1D B3GALT6 MIR200A MIR200B FAM132A SDF4 UBE2J2 PUSL1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-gkxk9:80/100.66.77.77:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline