This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01485474_cn
See other LOC105370757 CNV Assays ›
Gene Symbol
LOC105370757
Assay Reference Genome
Location

Chr.15:32670063 on build GRCh38
Cytoband
15q13.3
Assay ID Hs01485474_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

105370757

Gene Name:

uncharacterized LOC105370757

Gene Aliases:

-

Location:

Chr.15:32665702-32673065 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC105370757 NR_135505.1
BX088994.1

Target Gene Details

Entrez Gene ID:

6447

Gene Name:

secretogranin V

Gene Aliases:

7B2, P7B2, SGNE1, SgV

Location:

Chr.15:32641669-32697097 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SCG5 NM_001144757.1 NP_001138229.1
NM_003020.3 NP_003011.1
AK314965.1
BC005349.1 AAH05349.1
BC093053.1 AAH93053.1
BP377248.1
CR541824.1 CAG46623.1
Y00757.1 CAA68726.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv456785 Chr.15:32633875 - 33060788 on Build GRCh38 Gain FMN1 LOC105370757 LOC100131315 SCG5 ARHGAP11A GREM1
nsv428302 Chr.15:31646275 - 32694378 on Build GRCh38 Gain+Loss LOC105370757 ULK4P1 WHAMMP1 SCG5 LOC100653133 CHRNA7 ARHGAP11A LOC105370940 LOC101060588 OTUD7A LOC100996255 GOLGA8O GOLGA8K LOC101928042 GOLGA8N
esv2760023 Chr.15:31474097 - 32694378 on Build GRCh38 Gain+Loss LOC105370757 ULK4P1 WHAMMP1 SCG5 LOC100653133 CHRNA7 ARHGAP11A LOC105370940 LOC101060588 OTUD7A LOC100996255 GOLGA8O GOLGA8K LOC101928042 GOLGA8N
nsv1052620 Chr.15:32612402 - 34538731 on Build GRCh38 Gain EMC4 LOC105370757 KATNBL1 GOLGA8B AVEN SCG5 GOLGA8A NUTM1 ARHGAP11A MIR1233-2 TMCO5B NOP10 FMN1 MIR1233-1 LOC101928134 LOC100131315 PGBD4 CHRM5 LOC101928042 SLC12A6 RYR3 EMC7 LPCAT4 GREM1
nsv483039 Chr.15:32574635 - 32751321 on Build GRCh38 Gain LOC105370757 LOC100996255 LOC100131315 SCG5 LOC101928042 ARHGAP11A GREM1 GOLGA8N
esv3892661 Chr.15:32393132 - 32776080 on Build GRCh38 Gain LOC105370757 ULK4P1 WHAMMP1 SCG5 LOC100653133 ARHGAP11A LOC101060588 FMN1 LOC100996255 LOC100131315 GOLGA8O GOLGA8K LOC101928042 GREM1 GOLGA8N
esv33337 Chr.15:31689863 - 34575097 on Build GRCh38 Gain+Loss EMC4 LOC105370757 ULK4P1 WHAMMP1 GOLGA8B AVEN SCG5 NUTM1 MIR1233-2 MIR1233-1 LOC101928134 LOC100996255 LOC100131315 GOLGA8O LOC101928042 SLC12A6 RYR3 LPCAT4 GREM1 KATNBL1 LOC100653133 GOLGA8A CHRNA7 ARHGAP11A LOC105370940 LOC101060588 TMCO5B NOP10 FMN1 PGBD4 CHRM5 GOLGA8K EMC7 GOLGA8N
nsv469587 Chr.15:32529739 - 32694378 on Build GRCh38 Loss LOC105370757 WHAMMP1 LOC100996255 SCG5 LOC101928042 ARHGAP11A GOLGA8N
esv2422296 Chr.15:32506737 - 32929014 on Build GRCh38 Duplication FMN1 LOC105370757 WHAMMP1 LOC100996255 LOC100131315 SCG5 LOC101928042 ARHGAP11A GREM1 GOLGA8N
dgv4456n54 Chr.15:32604149 - 34557061 on Build GRCh38 Gain EMC4 LOC105370757 GOLGA8B AVEN SCG5 NUTM1 MIR1233-2 MIR1233-1 LOC101928134 LOC100131315 LOC101928042 SLC12A6 RYR3 LPCAT4 GREM1 KATNBL1 GOLGA8A ARHGAP11A TMCO5B NOP10 FMN1 PGBD4 CHRM5 EMC7 GOLGA8N

Back To Top

More Information


Set Membership:

Intragenic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
neuropeptide signaling pathway
peptide hormone processing
negative regulation of catalytic activity
regulation of hormone secretion
enzyme inhibitor activity
protein binding
GTP binding
unfolded protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-57b588f4f5-mk9w9:80/100.66.79.163:80.
git-commit: 377debcfccf3d85e21e16e6539a0b18561fa37ae
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.39.0-Offline