This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01497504_cn
See other SALL3 CNV Assays ›
Gene Symbol
SALL3
Assay Reference Genome
Location

Chr.18:78992165 on build GRCh38
Cytoband
18q23
Assay ID Hs01497504_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

27164

Gene Name:

spalt like transcription factor 3

Gene Aliases:

ZNF796

Location:

Chr.18:78980275-78998969 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SALL3 NM_171999.3 2 174 NP_741996.2
BC148296.2 2 174

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv577821 Chr.18:78980085 - 78994367 on Build GRCh38 Loss SALL3
nsv577793 Chr.18:78951024 - 79022312 on Build GRCh38 Loss SALL3
nsv828333 Chr.18:78955949 - 79017302 on Build GRCh38 Loss SALL3
nsv577794 Chr.18:78951366 - 79472709 on Build GRCh38 Gain SALL3 LOC107985149 LOC100996274 ATP9B NFATC1
nsv577795 Chr.18:78972221 - 79050030 on Build GRCh38 Loss SALL3
nsv2377 Chr.18:78991267 - 79025451 on Build GRCh38 Insertion SALL3
dgv564n67 Chr.18:78952775 - 79013936 on Build GRCh38 Gain SALL3
nsv1065902 Chr.18:78538045 - 80257174 on Build GRCh38 Gain RBFADN CTDP1 LOC105376875 ATP9B PARD6G-AS1 SALL3 LOC107985149 LOC100996274 ADNP2 HSBP1L1 TXNL4A LOC284241 RBFA PQLC1 KCNG2 NFATC1 PARD6G LOC284240
nsv828338 Chr.18:78963814 - 79001729 on Build GRCh38 Gain SALL3
esv2717497 Chr.18:78170182 - 79635788 on Build GRCh38 Deletion SALL3 LOC107985149 LOC100996274 ATP9B NFATC1 LOC284240
nsv1141945 Chr.18:78170394 - 79635820 on Build GRCh38 Deletion SALL3 LOC107985149 LOC100996274 ATP9B NFATC1 LOC284240
dgv565n67 Chr.18:78974294 - 78993921 on Build GRCh38 Gain SALL3

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
olfactory bulb interneuron development
neurogenesis
forelimb morphogenesis
hindlimb morphogenesis
negative regulation of smoothened signaling pathway
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mfdv2:80/100.66.77.127:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0