This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01513694_cn
See other NUDT22 CNV Assays ›
Gene Symbol
NUDT22
Assay Reference Genome
Location

Chr.11:64229940 on build GRCh38
Cytoband
11q13.1
Assay ID Hs01513694_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

84304

Gene Name:

nudix hydrolase 22

Gene Aliases:

-

Location:

Chr.11:64226258-64230016 on Build GRCh38

Assay Gene Location:

Within Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NUDT22 NM_001128612.2 6 920 NP_001122084.1
NM_001128613.2 5 935 NP_001122085.1
NM_001271831.1 5 821 NP_001258760.1
NM_032344.3 6 1034 NP_115720.1
BC006129.1 6 1018 AAH06129.1
BM923060.1 5 820

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1159904 Chr.11:64150129 - 64279169 on Build GRCh38 Deletion FKBP2 DNAJC4 LOC105369340 BAD FERMT3 PLCB3 NUDT22 TRPT1 STIP1 GPR137 VEGFB MACROD1 PPP1R14B
esv992196 Chr.11:64225062 - 64234883 on Build GRCh38 Deletion NUDT22 TRPT1 DNAJC4 VEGFB
nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion FKBP2 MIR7155 DNAJC4 COX8A PLCB3 ESRRA NUDT22 OTUB1 TRMT112 NAA40 PPP1R14B CCDC88B LOC105369340 BAD FERMT3 KCNK4-TEX40 TEX40 TRPT1 STIP1 GPR137 VEGFB MACROD1 FLRT1 PRDX5 KCNK4
dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss FKBP2 DNAJC4 LOC105369340 BAD FERMT3 PLCB3 ESRRA NUDT22 KCNK4-TEX40 TEX40 TRPT1 STIP1 GPR137 VEGFB TRMT112 PPP1R14B PRDX5 KCNK4
nsv555187 Chr.11:64185649 - 64260416 on Build GRCh38 Loss FKBP2 NUDT22 TRPT1 STIP1 DNAJC4 VEGFB LOC105369340 FERMT3 PPP1R14B PLCB3
nsv555184 Chr.11:64156974 - 64275525 on Build GRCh38 Loss FKBP2 DNAJC4 LOC105369340 BAD FERMT3 PLCB3 NUDT22 TRPT1 STIP1 GPR137 VEGFB MACROD1 PPP1R14B
nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss FKBP2 CCDC88B MIR7155 DNAJC4 LOC105369340 BAD FERMT3 PLCB3 ESRRA NUDT22 KCNK4-TEX40 TEX40 TRPT1 GPR137 VEGFB RPS6KA4 TRMT112 PPP1R14B MIR1237 PRDX5 KCNK4
nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss FKBP2 DNAJC4 LOC105369340 BAD FERMT3 PLCB3 ESRRA NUDT22 KCNK4-TEX40 TEX40 TRPT1 STIP1 GPR137 VEGFB TRMT112 MACROD1 FLRT1 PPP1R14B PRDX5 KCNK4

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Process(es)

protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline