This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01522326_cn
See other UCHL5 CNV Assays ›
Gene Symbol
UCHL5
Assay Reference Genome
Location

Chr.1:193029235 on build GRCh38
Cytoband
1q31.2
Assay ID Hs01522326_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

51377

Gene Name:

ubiquitin C-terminal hydrolase L5

Gene Aliases:

CGI-70, INO80R, UCH-L5, UCH37

Location:

Chr.1:193012366-193060140 on Build GRCh38

Assay Gene Location:

Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UCHL5 NM_001199261.1 6 642 NP_001186190.1
NM_001199262.1 6 642 NP_001186191.1
NM_001199263.1 6 642 NP_001186192.1
NM_015984.3 6 642 NP_057068.1
NR_037607.1 6 908
XM_005245244.4 6 725 XP_005245301.1
XM_005245246.4 6 725 XP_005245303.1
XM_005245247.4 6 577 XP_005245304.1
XM_005245248.4 6 600 XP_005245305.1
XM_005245249.4 6 496 XP_005245306.1
XM_006711366.3 6 725 XP_006711429.1
XM_006711367.3 6 725 XP_006711430.1
XM_006711368.3 6 725 XP_006711431.1
XM_006711369.2 6 725 XP_006711432.1
XM_006711370.3 6 678 XP_006711433.3
XM_006711371.3 6 496 XP_006711434.1
XM_011509604.1 6 725 XP_011507906.1
XM_011509607.2 6 536 XP_011507909.1
XM_011509608.2 7 897 XP_011507910.1
XM_011509609.2 7 801 XP_011507911.1
XM_017001429.1 6 725 XP_016856918.1
XM_017001430.1 6 725 XP_016856919.1
XM_017001431.1 6 725 XP_016856920.1
XM_017001432.1 6 725 XP_016856921.1
XM_017001433.1 6 725 XP_016856922.1
XM_017001434.1 6 678 XP_016856923.1
XM_017001435.1 6 678 XP_016856924.1
XM_017001436.1 7 897 XP_016856925.1
XM_017001437.1 7 897 XP_016856926.1
XM_017001438.1 6 520 XP_016856927.1
XM_017001439.1 6 941 XP_016856928.1
XM_017001440.1 6 905 XP_016856929.1
XM_017001441.1 6 881 XP_016856930.1
XM_017001442.1 6 496 XP_016856931.1
XM_017001443.1 6 496 XP_016856932.1
XM_017001444.1 6 890 XP_016856933.1
AF147717.1 6 640 AAD31528.1
AF151828.1 6 522 AAD34065.1
AF157320.1 6 657 AAF67486.1
AK024751.1 6 588
AK225794.1 6 575
AK296487.1
AK301381.1 6 908
AK314445.1 6 586
AK316064.1 5 480
BC015521.1 6 575 AAH15521.1
BC025369.2 6 620 AAH25369.1
BT006790.1 6 509 AAP35436.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv498n100 Chr.1:192992568 - 193114915 on Build GRCh38 Loss GLRX2 TROVE2 UCHL5 SCARNA18B
nsv466994 Chr.1:192394185 - 193552205 on Build GRCh38 Loss CDC73 MIR1278 SCARNA18B B3GALT2 GLRX2 TROVE2 RGS1 LINC01031 UCHL5 LINC01032 RGS13 MIR4426 RGS2
nsv3966 Chr.1:193007327 - 193035756 on Build GRCh38 Deletion UCHL5

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

cysteine protease

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
DNA recombination
transcription, DNA-templated
regulation of transcription, DNA-templated
ubiquitin-dependent protein catabolic process
negative regulation of endopeptidase activity
protein deubiquitination
lateral ventricle development
midbrain development
forebrain morphogenesis
regulation of proteasomal protein catabolic process
thiol-dependent ubiquitin-specific protease activity
endopeptidase inhibitor activity
protein binding
poly(A) RNA binding
proteasome binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0