This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01539403_cn
See other SLC2A4RG CNV Assays ›
Gene Symbol
SLC2A4RG
Assay Reference Genome
Location

Chr.20:63742434 on build GRCh38
Cytoband
20q13.33
Assay ID Hs01539403_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

56731

Gene Name:

SLC2A4 regulator

Gene Aliases:

GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19

Location:

Chr.20:63739858-63744050 on Build GRCh38

Assay Gene Location:

Within Exon 6
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC2A4RG NM_020062.3 6 834 NP_064446.2
AB044777.1 5 725 BAE71373.1
AB044786.1 4 395 BAD29732.1
AB052777.1 6 878 BAD29733.1
AF249267.3 6 779 AAF97516.2
AK298304.1 4 757
BC001402.2 5 605 AAH01402.2
BC017446.1 3 428 AAH17446.1
BC028349.2 AAH28349.2
BC052306.1 6 879 AAH52306.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion SLC2A4RG ZGPAT LIME1 ZBTB46
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion SLC2A4RG TNFRSF6B ARFRP1 ZGPAT RTEL1-TNFRSF6B STMN3 RTEL1 LIME1 ZBTB46-AS1 ZBTB46
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion SLC2A4RG TNFRSF6B ARFRP1 ZGPAT RTEL1-TNFRSF6B STMN3 RTEL1 LIME1 ZBTB46
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss MIR941-1 TCEA2 TNFRSF6B OPRL1 UCKL1-AS1 PCMTD2 MIR941-4 DNAJC5 C20orf181 NPBWR2 SLC2A4RG LINC00176 ZNF512B MYT1 LIME1 SOX18 LKAAEAR1 UCKL1 ARFRP1 TPD52L2 MIR6813 STMN3 RTEL1 ZBTB46-AS1 PRPF6 ABHD16B LINC00266-1 GMEB2 ZGPAT RTEL1-TNFRSF6B MIR941-3 MIR941-5 SAMD10 LOC100505771 MIR941-2 MIR647 RGS19 MIR1914 ZBTB46
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss SLC2A4RG ZGPAT LIME1 ZBTB46
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain SLC2A4RG ZGPAT LIME1 ZBTB46
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss TNFRSF6B ARFRP1 STMN3 RTEL1 FNDC11 PTK6 GMEB2 SLC2A4RG HELZ2 ZGPAT RTEL1-TNFRSF6B SRMS LIME1 LOC100505771 EEF1A2 ZBTB46 PPDPF
dgv7684n54 Chr.20:63738644 - 63789659 on Build GRCh38 Loss SLC2A4RG LIME1 ZBTB46
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss TNFRSF6B ARFRP1 STMN3 RTEL1 FNDC11 PTK6 KCNQ2 GMEB2 SLC2A4RG HELZ2 ZGPAT RTEL1-TNFRSF6B SRMS LIME1 LOC100505771 EEF1A2 ZBTB46 PPDPF
dgv7680n54 Chr.20:63737451 - 63743088 on Build GRCh38 Gain SLC2A4RG LIME1

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline