This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01578412_cn
See other NR1D1 CNV Assays ›
Gene Symbol
NR1D1
Assay Reference Genome
Location

Chr.17:40093902 on build GRCh38
Cytoband
17q21.1
Assay ID Hs01578412_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9572

Gene Name:

nuclear receptor subfamily 1 group D member 1

Gene Aliases:

EAR1, THRA1, THRAL, ear-1, hRev

Location:

Chr.17:40092784-40100725 on Build GRCh38

Assay Gene Location:

Overlaps Intron 7 - Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NR1D1 NM_021724.4 NP_068370.1
AK300155.1
AK315270.1
BC047875.1
BC056148.1 AAH56148.1
BM671861.1
HQ692861.1
M24898.1 AAA52335.1
X72631.1 CAB53540.1

Target Gene Details

Entrez Gene ID:

7067

Gene Name:

thyroid hormone receptor, alpha

Gene Aliases:

AR7, CHNG6, EAR7, ERB-T-1, ERBA, ERBA1, NR1A1, THRA1, THRA2, c-ERBA-1

Location:

Chr.17:40062193-40093867 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 11
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
THRA NM_001190918.1 NP_001177847.1
NM_001190919.1 NP_001177848.1
NM_003250.5 NP_003241.2
BC000261.1 AAH00261.1
BC002728.2 AAH02728.1
BC035137.1 AAH35137.1
M24900.1 1 393 AAA52333.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv833444 Chr.17:40086570 - 40281028 on Build GRCh38 Loss MSL1 CASC3 NR1D1 WIPF2 RAPGEFL1 THRA MIR6866 MIR6867

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

C4 zinc finger nuclear receptor DNA-binding transcription factor zinc finger transcription factor gene-specific transcriptional regulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
glycogen biosynthetic process
transcription initiation from RNA polymerase II promoter
circadian rhythm
proteasomal protein catabolic process
negative regulation of receptor biosynthetic process
regulation of lipid metabolic process
cell differentiation
intracellular receptor signaling pathway
circadian regulation of gene expression
negative regulation of toll-like receptor 4 signaling pathway
regulation of gluconeogenesis by regulation of transcription from RNA polymerase II promoter
regulation of circadian rhythm
steroid hormone mediated signaling pathway
response to leptin
regulation of fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
circadian temperature homeostasis
regulation of insulin secretion involved in cellular response to glucose stimulus
regulation of type B pancreatic cell proliferation
positive regulation of bile acid biosynthetic process
cellular response to lipopolysaccharide
regulation of cholesterol homeostasis
positive regulation of cholesterol homeostasis
cartilage condensation
ossification
kidney development
liver development
regulation of thyroid hormone mediated signaling pathway
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
brain development
learning or memory
regulation of heart contraction
female courtship behavior
response to cold
hormone-mediated signaling pathway
positive regulation of myotube differentiation
negative regulation of RNA polymerase II transcriptional preinitiation complex assembly
erythrocyte differentiation
adrenal gland development
thyroid gland development
response to nutrient levels
regulation of myeloid cell apoptotic process
response to drug
cytoplasmic sequestering of transcription factor
negative regulation of sequence-specific DNA binding transcription factor activity
positive regulation of female receptivity
positive regulation of transcription from RNA polymerase II promoter
digestive tract development
embryonic organ development
regulation of lipid catabolic process
Type I pneumocyte differentiation
negative regulation of DNA-templated transcription, initiation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
core promoter sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription corepressor binding
steroid hormone receptor activity
transcription corepressor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
protein binding
zinc ion binding
heme binding
transcription regulatory region DNA binding
transcription regulatory region sequence-specific DNA binding
steroid receptor RNA activator RNA binding
transcription factor activity, sequence-specific DNA binding
transcription factor activity, RNA polymerase II distal enhancer sequence-specific binding
thyroid hormone receptor activity
transcription factor binding
TBP-class protein binding
protein domain specific binding
chromatin DNA binding
protein complex binding
protein homodimerization activity
intronic transcription regulatory region DNA binding
protein heterodimerization activity
thyroid hormone binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-7669c5b596-qmnfm:80/100.66.79.90:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0