This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01586783_cn
See other LOC100996517 CNV Assays ›
Gene Symbol
LOC100996517
Assay Reference Genome
Location

Chr.1:120163281 on build GRCh38
Cytoband
1p12
Assay ID Hs01586783_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

100996517

Gene Name:

vesicle-trafficking protein SEC22b

Gene Aliases:

-

Location:

Chr.1:148766340-148786183 on Build GRCh38

Assay Gene Location:

Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100996517 XM_017003060.1 3 760 XP_016858549.1

Target Gene Details

Entrez Gene ID:

100996716

Gene Name:

vesicle-trafficking protein SEC22b

Gene Aliases:

CH17-189H20.1

Location:

Chr.1:149175714-149363915 on Build GRCh38

Assay Gene Location:

Within Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100996716 XM_017003058.1 3 759 XP_016858547.1
XM_017003059.1 4 851 XP_016858548.1

Target Gene Details

Entrez Gene ID:

9554

Gene Name:

SEC22 homolog B, vesicle trafficking protein (gene/pseudogene)

Gene Aliases:

ERS-24, SEC22L1

Location:

Chr.1:120152520-120176534 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SEC22B NM_004892.5 3 415 NP_004883.3
XM_017002856.1 3 428 XP_016858345.1
XM_017002857.1 3 857 XP_016858346.1
AF047442.1 3 339 AAC39893.1
AK289875.1 3 409
BC001364.2 3 394 AAH01364.1
CD619408.1 3 445

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain LOC100505824 LOC100132057 ACP6 LINC00624 LINC01138 LOC100996763 NBPF11 LOC105371226 RNVU1-6 NBPF14 PRKAB2 LOC105371242 PDE4DIP LOC101927468 BCL9 PPIAL4G GJA5 NOTCH2NL GJA8 MIR5087 RNVU1-7 RNU1-13P CHD1L LOC284561 RNVU1-3 RNVU1-1 LOC101928979 NBPF13P NUDT4P2 MIR6077 NBPF10 GPR89B HYDIN2 PDZK1P1 PDIA3P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 FMO5 LOC100996517
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss RBM8A LINC01138 POLR3C NBPF11 LOC105371226 RNVU1-6 NBPF14 PDE4DIP PPIAL4G PEX11B GJA5 NOTCH2NL TXNIP PDZK1 GJA8 MIR5087 LOC284561 RNVU1-1 MIR6736 ITGA10 NBPF13P NUDT4P2 HFE2 GNRHR2 MIR6077 NBPF10 GPR89B HYDIN2 PDZK1P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 ANKRD34A FMO5 CD160 LOC100505824 LIX1L LOC100132057 ACP6 NUDT17 LINC00624 LOC100996763 PRKAB2 LOC105371242 LOC101927468 BCL9 RNVU1-7 RNU1-13P CHD1L RNVU1-3 ANKRD35 LOC101928979 GPR89A POLR3GL PDIA3P1 PIAS3 LOC100996517 RNF115
dgv326n100 Chr.1:148774675 - 148889555 on Build GRCh38 Gain LOC100996517 PDE4DIP
esv2581967 Chr.1:148768990 - 148790234 on Build GRCh38 Gain LOC100996517
dgv324n100 Chr.1:148736158 - 148934526 on Build GRCh38 Gain NUDT4P1 LOC100996517 PDE4DIP
dgv17n16 Chr.1:148696576 - 148805441 on Build GRCh38 Deletion NUDT4P1 LOC100996763 LOC100996517
nsv946952 Chr.1:148577301 - 149056658 on Build GRCh38 Duplication NBPF9 NUDT4P1 LOC100996763 LOC100996517 NBPF14 PDE4DIP
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication LOC100505824 LOC100132057 ACP6 LINC00624 LINC01138 LOC100996763 NBPF11 LOC105371226 NBPF14 PRKAB2 LOC105371242 PDE4DIP LOC101927468 BCL9 PPIAL4G GJA5 NOTCH2NL GJA8 MIR5087 RNVU1-7 RNU1-13P CHD1L LOC284561 RNVU1-3 RNVU1-1 NBPF13P NUDT4P2 MIR6077 GPR89B HYDIN2 PDZK1P1 PDIA3P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 FMO5 LOC100996517
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain LOC100505824 LOC100132057 ACP6 LINC00624 LINC01138 LOC100996763 NBPF11 LOC105371226 RNVU1-6 NBPF14 PRKAB2 LOC105371242 PDE4DIP LOC101927468 BCL9 PPIAL4G GJA5 NOTCH2NL GJA8 MIR5087 RNVU1-7 RNU1-13P CHD1L LOC284561 RNVU1-3 RNVU1-1 LOC101928979 NBPF13P NUDT4P2 MIR6077 NBPF10 GPR89B HYDIN2 PDZK1P1 PDIA3P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 FMO5 LOC100996517
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion LOC100505824 LOC100132057 ACP6 LINC00624 LINC01138 LOC100996763 NBPF11 LOC105371226 NBPF14 PRKAB2 LOC105371242 PDE4DIP LOC101927468 BCL9 PPIAL4G GJA5 NBPF9 NOTCH2NL GJA8 MIR5087 RNVU1-7 RNU1-13P CHD1L LOC284561 RNVU1-3 RNVU1-1 LOC101928979 NBPF13P NUDT4P2 MIR6077 NBPF10 GPR89B HYDIN2 PDZK1P1 PDIA3P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 FMO5 LOC100996517
dgv328n100 Chr.1:148598832 - 148804379 on Build GRCh38 Gain NUDT4P1 LOC100996763 LOC100996517 NBPF14
nsv1077297 Chr.1:148761929 - 148791400 on Build GRCh38 Duplication LOC100996517
nsv1141148 Chr.1:148761729 - 148791399 on Build GRCh38 Duplication LOC100996517
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss LOC100505824 LOC100132057 ACP6 LINC00624 LINC01138 LOC100996763 NBPF11 LOC105371226 NBPF14 PRKAB2 LOC105371242 PDE4DIP LOC101927468 BCL9 PPIAL4G GJA5 NOTCH2NL GJA8 MIR5087 RNVU1-7 RNU1-13P CHD1L LOC284561 RNVU1-3 RNVU1-1 NBPF13P NUDT4P2 MIR6077 NBPF10 GPR89B HYDIN2 PDZK1P1 PDIA3P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 FMO5 LOC100996517
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion LOC100505824 LIX1L LOC100132057 ACP6 LINC00624 LINC01138 LOC100996763 NBPF11 LOC105371226 RNVU1-6 NBPF14 PRKAB2 LOC105371242 PDE4DIP LOC101927468 BCL9 PPIAL4G GJA5 NOTCH2NL TXNIP GJA8 MIR5087 RNVU1-7 RNU1-13P CHD1L LOC284561 RNVU1-3 RNVU1-1 LOC101928979 NBPF13P NUDT4P2 HFE2 MIR6077 NBPF10 GPR89B HYDIN2 PDZK1P1 POLR3GL PDIA3P1 NUDT4P1 LOC728989 NBPF12 CH17-408M7.1 RNVU1-8 ANKRD34A FMO5 LOC100996517
nsv946254 Chr.1:148752296 - 148791404 on Build GRCh38 Duplication NUDT4P1 LOC100996517
nsv1005230 Chr.1:148737400 - 149018844 on Build GRCh38 Gain NUDT4P1 LOC100996517 PDE4DIP

Back To Top

More Information


Additional Information:

This assay has 4 genomic targets in NCBI build GRCh38

Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
protein transport
regulation of organelle organization
positive regulation of protein catabolic process
COPII vesicle coating
vesicle fusion with Golgi apparatus
SNARE binding
SNAP receptor activity
protein binding
syntaxin binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0