This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01591205_cn
See other VSTM1 CNV Assays ›
Gene Symbol
VSTM1
Assay Reference Genome
Location

Chr.19:54063769 on build GRCh38
Cytoband
19q13.42
Assay ID Hs01591205_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284415

Gene Name:

V-set and transmembrane domain containing 1

Gene Aliases:

SIRL-1, SIRL1, UNQ3033

Location:

Chr.19:54040825-54063966 on Build GRCh38

Assay Gene Location:

Overlaps Intron 1 - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
VSTM1 NM_001288791.1 NP_001275720.1
NM_001288792.1 NP_001275721.1
NM_001288793.1 NP_001275722.1
NM_198481.3 NP_940883.2
NR_110142.1
XM_011526845.1 XP_011525147.1
XM_011526846.1 XP_011525148.1
XM_011526847.1 XP_011525149.1
XM_011526848.1 XP_011525150.1
XM_011526849.1 XP_011525151.1
XM_017026666.1 XP_016882155.1
AY358542.1 AAQ88906.1
BC100943.2 AAI00944.1
DQ479397.1
FJ584316.1
FJ584317.1
FJ584318.1
FJ584319.1
FJ584320.1
FJ882051.1
FN398145.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv458782 Chr.19:53961144 - 54093354 on Build GRCh38 Gain MIR935 CACNG8 CACNG6 TARM1 VSTM1
nsv510773 Chr.19:54017180 - 54132236 on Build GRCh38 Deletion OSCAR TFPT NDUFA3 TARM1 PRPF31 VSTM1
nsv182 Chr.19:54050737 - 54097924 on Build GRCh38 Deletion OSCAR TARM1 VSTM1
nsv1063127 Chr.19:53926876 - 54068977 on Build GRCh38 Gain MIR935 CACNG8 CACNG6 VSTM1 CACNG7
esv2718812 Chr.19:53184636 - 54322450 on Build GRCh38 Deletion CNOT3 LILRA6 MIR518E ZNF845 MIR518F MIR525 MIR518B MIR4752 MIR522 MIR520B MIR519D MIR521-1 FAM90A27P MIR371B ZNF818P TARM1 PRPF31 TSEN34 MIR519A1 MIR527 MIR516B1 ZNF813 MIR935 LOC105372457 MIR520C MIR517C ZNF665 MIR516A2 VN1R2 NDUFA3 MIR518A1 MIR520H LILRB2 NLRP12 MIR1323 RPS9 ZNF331 MIR524 MIR526A1 DPRX MIR512-1 MIR519C MIR515-1 CACNG8 LILRA5 MIR373 VN1R4 MIR516A1 MIR520F LILRB5 MIR1283-2 MIR526A2 MIR498 TFPT MIR515-2 MIR1283-1 MYADM BIRC8 MIR517A MIR519B ZNF761 MIR526B MIR518A2 ZNF677 MIR518D OSCAR MIR372 LENG1 MIR371A MIR520E MIR519E MIR520D ZNF525 MIR520A MBOAT7 ZNF765 VSTM1 MIR521-2 LILRB3 TMC4 TPM3P9 MIR523 MIR517B MIR512-2 MIR516B2 MIR520G CACNG6 PRKCG LOC284379 MIR518C MIR519A2 CACNG7
esv3644780 Chr.19:54057724 - 54066399 on Build GRCh38 Gain VSTM1
nsv833877 Chr.19:53951529 - 54117323 on Build GRCh38 Loss MIR935 OSCAR TFPT CACNG8 CACNG6 NDUFA3 TARM1 PRPF31 VSTM1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

immune system process
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline