This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01604883_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1312855 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01604883_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,505.00
Your Price
Online offer:
4505.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Within Exon 14
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 14 1778 NP_001243385.1
NM_001256460.1 13 1412 NP_001243389.1
NM_001256462.1 10 1011 NP_001243391.1
NM_001256463.1 10 1002 NP_001243392.1
NM_017871.5 12 1305 NP_060341.2
XM_011541647.1 14 1456 XP_011539949.1
XM_011541648.1 14 1460 XP_011539950.1
XM_011541650.1 12 1185 XP_011539952.1
XM_017001557.1 13 1261 XP_016857046.1
XM_017001558.1 14 1315 XP_016857047.1
AK000549.1 4 449 BAA91246.1
AK021939.1 10 968 BAB13943.1
AK023356.1 12 1223 BAB14541.1
AK056652.1 14 1700
AK058018.1 10 977
AK092190.1 9 1038
AK123641.1 3 2092
AK291387.1 14 1741
AK297350.1 13 1362
AL136813.1 12 1284 CAB66747.1
BC000675.2 9 845 AAH00675.2
BC007978.2 12 1267 AAH07978.1
BC008041.2 AAH08041.1
BC013904.2 8 714 AAH13904.2
BC020199.1 AAH20199.1
CR533557.1 12 1226 CAG38588.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC100134822 AURKAIP1 LOC100287934 MTND2P28 MIR200B HES4 LOC100506504 LOC107984841 LOC729737 MIR200A SCNN1D MIR6726 LOC105378947 SLC35E2 LOC148413 TTLL10-AS1 MIR1302-2 ISG15 MIR6727 TMEM88B TAS1R3 LOC102724312 KLHL17 LOC100132287 LOC284600 ATAD3C B3GALT6 RNF223 LINC01342 LOC107985729 DDX11L1 PLEKHN1 LINC01128 SLC35E2B ATAD3B CDK11B GNB1 CPTP SAMD11 LOC105378948 OR4F16 DVL1 TMEM52 SKI PUSL1 MMP23A MIR6723 CFAP74 NOC2L FAM138A FAAP20 OR4F29 SSU72 ANKRD65 C1orf159 LOC100288175 FNDC10 PERM1 LOC107985728 CDK11A FAM41C LOC105378949 WASH7P ACAP3 LOC100129534 LOC100288069 MIR429 MORN1 MRPL20 SDF4 TTLL10 MMP23B ATAD3A MIR6859-1 MTND1P23 LOC105378589 TNFRSF4 PRKCZ LOC105378592 LOC100130417 MIB2 LOC100133331 MXRA8 AGRN VWA1 LINC00115 LOC101928626 UBE2J2 OR4F5 TMEM240 GABRD FAM132A CPSF3L LOC105378591 MIR6859-2 MIR6808 NADK CALML6 FAM87B CCNL2 TNFRSF18 LOC102725121
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss DVL1 CPTP MIR6726 MIR6808 MIR6727 TAS1R3 PUSL1 ACAP3 CPSF3L
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss SCNN1D DVL1 CPTP MIR6726 MIR6808 MIR6727 MXRA8 TAS1R3 AURKAIP1 PUSL1 ACAP3 CPSF3L
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain DVL1 CPTP MIR6808 MIR6727 MXRA8 TAS1R3 AURKAIP1 PUSL1 ACAP3 CPSF3L
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss SDF4 B3GALT6 AURKAIP1 RNF223 LINC01342 TTLL10 TNFRSF4 MIR200B C1orf159 MIR200A SCNN1D CPTP MIR6726 MXRA8 UBE2J2 TTLL10-AS1 FAM132A CPSF3L DVL1 MIR6808 MIR6727 TAS1R3 PUSL1 ACAP3 TNFRSF18 MIR429
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss DVL1 CPTP MIR6808 MIR6727 MXRA8 TAS1R3 PUSL1 ACAP3 CPSF3L
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion SCNN1D DVL1 CPTP MIR6726 MIR6808 MIR6727 MXRA8 TAS1R3 AURKAIP1 PUSL1 ACAP3 CPSF3L
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication SDF4 B3GALT6 RNF223 LINC01342 TTLL10 TNFRSF4 MIR200B C1orf159 LOC100288175 MIR200A SCNN1D CPTP MIR6726 AGRN LOC105378948 UBE2J2 TTLL10-AS1 FAM132A CPSF3L MIR6727 TAS1R3 PUSL1 ACAP3 TNFRSF18 MIR429
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss AURKAIP1 SSU72 LOC100287934 ANKRD65 MIR200B HES4 C1orf159 LOC100288175 MIR200A SCNN1D FNDC10 PERM1 LOC107985728 MIR6726 CDK11A FAM41C SLC35E2 LOC148413 TTLL10-AS1 ISG15 MIR6727 TMEM88B TAS1R3 LOC102724312 KLHL17 ACAP3 LOC100288069 MIR429 MRPL20 LOC284600 SDF4 ATAD3C B3GALT6 RNF223 LINC01342 TTLL10 MMP23B ATAD3A TNFRSF4 LOC107985729 PLEKHN1 LINC01128 LOC100130417 SLC35E2B ATAD3B MIB2 CDK11B CPTP SAMD11 MXRA8 AGRN VWA1 LINC00115 LOC105378948 UBE2J2 TMEM240 FAM132A CPSF3L DVL1 MIR6808 NADK PUSL1 MMP23A FAM87B CCNL2 NOC2L TNFRSF18
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss DVL1 CPTP MIR6808 MIR6727 MXRA8 TAS1R3 PUSL1 ACAP3 CPSF3L
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain SCNN1D DVL1 CPTP MIR6726 MIR6808 MIR6727 MXRA8 TAS1R3 PUSL1 ACAP3 CPSF3L
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MRPL20 SDF4 ATAD3C B3GALT6 AURKAIP1 LINC01342 TTLL10 MMP23B ATAD3A SSU72 TNFRSF4 LOC107985729 ANKRD65 MIR200B ATAD3B MIB2 C1orf159 CDK11B MIR200A SCNN1D FNDC10 CPTP MIR6726 MXRA8 VWA1 LOC148413 UBE2J2 TMEM240 TTLL10-AS1 FAM132A CPSF3L DVL1 MIR6808 MIR6727 TMEM88B TAS1R3 LOC102724312 PUSL1 ACAP3 CCNL2 TNFRSF18 MIR429
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss SCNN1D CPTP MIR6726 MXRA8 AURKAIP1 UBE2J2 CPSF3L DVL1 MIR6808 MIR6727 TAS1R3 PUSL1 ACAP3
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MRPL20 LOC284600 SDF4 B3GALT6 AURKAIP1 RNF223 LINC01342 TTLL10 TNFRSF4 PLEKHN1 ANKRD65 LOC100130417 MIR200B HES4 C1orf159 LOC100288175 MIR200A SCNN1D PERM1 CPTP LOC107985728 MIR6726 SAMD11 MXRA8 AGRN LOC148413 LOC105378948 UBE2J2 TTLL10-AS1 FAM132A CPSF3L DVL1 ISG15 MIR6808 MIR6727 TMEM88B TAS1R3 PUSL1 KLHL17 ACAP3 CCNL2 NOC2L TNFRSF18 MIR429
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR200A SCNN1D MIR6726 SDF4 B3GALT6 TTLL10 UBE2J2 TTLL10-AS1 FAM132A CPSF3L TNFRSF4 MIR6727 PUSL1 ACAP3 MIR200B TNFRSF18 MIR429

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0