This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01656667_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1625357 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01656667_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 7 1183 NP_001164157.1
NM_001170687.1 7 1141 NP_001164158.1
NM_001170688.1 6 988 NP_001164159.1
NM_001170689.1 6 963 NP_001164160.1
NM_080875.2 7 1183 NP_543151.2
NR_033183.1 7 966
XM_006710372.1 7 1155 XP_006710435.1
XM_011540731.2 7 1857 XP_011539033.1
XM_011540736.2 7 1857 XP_011539038.1
XM_011540737.2 7 1857 XP_011539039.1
XM_011540741.2 7 1857 XP_011539043.1
XM_011540742.2 7 1857 XP_011539044.1
XM_017000349.1 7 1857 XP_016855838.1
XM_017000350.1 7 1857 XP_016855839.1
XM_017000351.1 7 1839 XP_016855840.1
XM_017000352.1 7 1815 XP_016855841.1
XM_017000353.1 7 1857 XP_016855842.1
XM_017000354.1 7 1857 XP_016855843.1
XM_017000355.1 7 1857 XP_016855844.1
XM_017000356.1 7 1857 XP_016855845.1
XM_017000357.1 6 1662 XP_016855846.1
XM_017000358.1 7 1155 XP_016855847.1
XM_017000359.1 7 1155 XP_016855848.1
XM_017000360.1 7 1102 XP_016855849.1
XM_017000361.1 7 1155 XP_016855850.1
XM_017000362.1 7 1195 XP_016855851.1
XM_017000363.1 7 1158 XP_016855852.1
XM_017000364.1 6 960 XP_016855853.1
XM_017000365.1 7 885 XP_016855854.1
XM_017000366.1 7 885 XP_016855855.1
AB074480.1 7 1094 BAB92950.1
AB076691.1 7 1052 BAC00992.1
AB076692.1 6 899 BAC00993.1
AB076693.1 7 1094 BAC00994.1
AB097000.1 7 1017 BAC77353.1
AK091610.1 2 95 BAC03707.1
AK095914.1 BAC04646.1
AK096295.1 2 94 BAC04752.1
AK097106.1 4 518 BAC04952.1
AK098785.1 7 1052 BAC05413.1
AK122751.1 7 885
AK122863.1 7 1084
AK128167.1 6 923
AK128553.1 7 1101
AK293642.1 6 1163
AK302757.1 7 1135
AK309362.1 6 1161
AL834527.1 2 215 CAD39183.1
BC016490.1 7 1020 AAH16490.1
BC037542.1 7 1063 AAH37542.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion FNDC10 SLC35E2B MMP23A SLC35E2 MIB2 CDK11A CDK11B MMP23B GNB1 NADK
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss FNDC10 SLC35E2B MMP23A SLC35E2 SSU72 MIB2 CDK11A CDK11B MMP23B
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain SLC35E2B MMP23A MIB2 CDK11A CDK11B MMP23B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss AURKAIP1 TMEM88B LINC01128 MIR6727 CCNL2 CPTP CDK11A FAM132A PUSL1 MIR200B FAM87B LOC100287934 TNFRSF18 LOC148413 LOC100288069 ISG15 MRPL20 TMEM240 MIB2 MIR6726 B3GALT6 ATAD3C MMP23B VWA1 MIR429 MXRA8 CDK11B LOC100130417 TTLL10 ATAD3B LOC107985729 PLEKHN1 HES4 TAS1R3 DVL1 ACAP3 SLC35E2B LOC100288175 AGRN SAMD11 SSU72 ATAD3A TTLL10-AS1 TNFRSF4 LINC01342 LOC102724312 LOC284600 LOC105378948 MMP23A MIR6808 SCNN1D SDF4 UBE2J2 C1orf159 FNDC10 FAM41C LOC107985728 NOC2L PERM1 NADK RNF223 ANKRD65 KLHL17 SLC35E2 MIR200A CPSF3L LINC00115
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LINC01128 LOC100506504 CCNL2 PUSL1 GNB1 MIR200B FAM87B SKI DDX11L1 LOC100287934 LOC148413 ISG15 TMEM240 MIB2 MIR6859-1 ATAD3C MIR1302-2 LOC100129534 MXRA8 OR4F16 CDK11B LOC100130417 ATAD3B LOC107985729 PLEKHN1 ACAP3 LOC105378947 CFAP74 FAM138A LOC100288175 SAMD11 ATAD3A TNFRSF4 LOC105378589 LOC102724312 LOC729737 LOC105378948 MIR6859-2 SCNN1D SDF4 UBE2J2 LOC107984841 FAM41C LOC107985728 TMEM52 RNF223 ANKRD65 KLHL17 SLC35E2 FAAP20 CPSF3L LINC00115 LOC101928626 MORN1 LOC100134822 AURKAIP1 TMEM88B MIR6727 CPTP CDK11A FAM132A LOC100132287 TNFRSF18 LOC100288069 OR4F5 GABRD LOC105378592 MRPL20 MIR6726 B3GALT6 MMP23B VWA1 MIR429 TTLL10 HES4 TAS1R3 LOC105378591 DVL1 MIR6723 PRKCZ SLC35E2B AGRN SSU72 TTLL10-AS1 LINC01342 OR4F29 LOC284600 MMP23A MTND2P28 MIR6808 WASH7P LOC102725121 C1orf159 FNDC10 LOC105378949 NOC2L PERM1 NADK MTND1P23 CALML6 MIR200A LOC100133331
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication FNDC10 SLC35E2B MMP23A SLC35E2 SSU72 MIB2 CDK11A CDK11B MMP23B GNB1 NADK
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion AURKAIP1 TMEM88B MIR6727 SSU72 ATAD3A CCNL2 CPTP FAM132A PUSL1 TTLL10-AS1 MIR200B TNFRSF4 LINC01342 LOC102724312 MIR6808 SCNN1D TNFRSF18 SDF4 LOC148413 UBE2J2 C1orf159 FNDC10 MRPL20 TMEM240 MIB2 MIR6726 B3GALT6 ATAD3C MMP23B ANKRD65 VWA1 MIR429 MXRA8 CDK11B MIR200A TTLL10 ATAD3B LOC107985729 CPSF3L TAS1R3 DVL1 ACAP3
esv2661270 Chr.1:1625365 - 1636030 on Build GRCh38 Deletion MIB2 CDK11B MMP23B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss FNDC10 SLC35E2B MMP23A SLC35E2 SSU72 MIB2 CDK11A CDK11B MMP23B NADK
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss SLC35E2B MIB2 CDK11B MMP23B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss PRKCZ CFAP74 TMEM88B SLC35E2B SSU72 ATAD3A CCNL2 CDK11A GNB1 LOC105378589 LOC102724312 MMP23A LOC148413 GABRD FNDC10 MRPL20 LOC105378949 TMEM240 MIB2 TMEM52 ATAD3C MMP23B NADK ANKRD65 VWA1 SLC35E2 CALML6 CDK11B ATAD3B LOC107985729 LOC105378591

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0