This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01675842_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1256056 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01675842_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Overlaps Intron 9 - Exon 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK098803.1
AK291935.1
AL555293.3
AW411508.1
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BG499172.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication SCNN1D LINC01342 TTLL10 UBE2J2 TNFRSF18 PUSL1 MIR200A B3GALT6 CPSF3L TAS1R3 TNFRSF4 LOC105378948 CPTP RNF223 FAM132A MIR6726 MIR200B SDF4 MIR6727 LOC100288175 C1orf159 TTLL10-AS1 MIR429 ACAP3 AGRN
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss VWA1 SCNN1D MIB2 ATAD3C LINC01342 TTLL10 UBE2J2 TNFRSF18 MIR200A LINC00115 LOC100130417 SLC35E2 TAS1R3 DVL1 TNFRSF4 KLHL17 MXRA8 MIR6808 FAM87B FAM41C CPTP SAMD11 ANKRD65 CDK11A FAM132A CCNL2 AURKAIP1 MIR200B MIR6727 LOC100288175 C1orf159 NADK MMP23A FNDC10 NOC2L TTLL10-AS1 LOC107985729 LOC100287934 ATAD3B PUSL1 MMP23B PERM1 B3GALT6 CPSF3L MRPL20 SLC35E2B LINC01128 LOC105378948 LOC107985728 RNF223 HES4 PLEKHN1 MIR6726 ATAD3A LOC100288069 TMEM88B SSU72 SDF4 LOC102724312 CDK11B TMEM240 ISG15 LOC148413 MIR429 LOC284600 ACAP3 AGRN
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss VWA1 SCNN1D ATAD3C UBE2J2 TNFRSF18 GNB1 LOC105378949 DVL1 TNFRSF4 MXRA8 MIR6808 FAM87B MORN1 CPTP ANKRD65 FAAP20 CCNL2 GABRD WASH7P OR4F16 MIR6727 LOC100288175 NADK FNDC10 LOC105378589 LOC105378591 FAM138A LOC100134822 B3GALT6 CPSF3L MRPL20 SLC35E2B LINC01128 LOC100132287 LOC105378948 PLEKHN1 TMEM88B SDF4 PRKCZ LOC102724312 LOC105378592 CDK11B ISG15 OR4F5 LOC100506504 LOC105378947 MIR429 LOC284600 ACAP3 MIB2 SKI LINC01342 TTLL10 LOC100133331 MIR200A LINC00115 LOC100130417 MIR6859-1 SLC35E2 TAS1R3 DDX11L1 KLHL17 FAM41C SAMD11 MIR1302-2 LOC102725121 CDK11A MIR6859-2 FAM132A LOC107984841 AURKAIP1 MIR200B C1orf159 CFAP74 MMP23A MIR6723 NOC2L TTLL10-AS1 LOC107985729 LOC100287934 ATAD3B PUSL1 MMP23B CALML6 PERM1 LOC101928626 LOC729737 MTND2P28 TMEM52 LOC107985728 RNF223 HES4 LOC100129534 MIR6726 ATAD3A OR4F29 LOC100288069 MTND1P23 SSU72 TMEM240 LOC148413 AGRN
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss SCNN1D FAM132A MIR6726 UBE2J2 B3GALT6 SDF4 ACAP3
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LINC01342 TTLL10 UBE2J2 TNFRSF18 MIR200A LINC00115 PERM1 B3GALT6 LOC100130417 LINC01128 TNFRSF4 KLHL17 FAM87B FAM41C LOC105378948 LOC107985728 SAMD11 RNF223 HES4 PLEKHN1 FAM132A MIR200B SDF4 LOC100288175 C1orf159 ISG15 NOC2L TTLL10-AS1 MIR429 LOC284600 AGRN
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion VWA1 SCNN1D MIB2 ATAD3C LINC01342 TTLL10 ATAD3B UBE2J2 TNFRSF18 PUSL1 MIR200A MMP23B B3GALT6 CPSF3L MRPL20 TAS1R3 DVL1 TNFRSF4 MXRA8 MIR6808 CPTP ANKRD65 FAM132A CCNL2 MIR6726 ATAD3A AURKAIP1 TMEM88B SSU72 MIR200B SDF4 MIR6727 LOC102724312 CDK11B C1orf159 TMEM240 LOC148413 FNDC10 TTLL10-AS1 LOC107985729 MIR429 ACAP3
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain SCNN1D LINC01342 TTLL10 UBE2J2 TNFRSF18 PUSL1 MIR200A PERM1 B3GALT6 LOC100130417 CPSF3L MRPL20 TAS1R3 DVL1 TNFRSF4 KLHL17 MXRA8 MIR6808 LOC105378948 LOC107985728 CPTP SAMD11 RNF223 HES4 ANKRD65 PLEKHN1 FAM132A CCNL2 MIR6726 AURKAIP1 TMEM88B MIR200B SDF4 MIR6727 LOC100288175 C1orf159 ISG15 LOC148413 NOC2L TTLL10-AS1 MIR429 LOC284600 ACAP3 AGRN
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain SCNN1D FAM132A LINC01342 TTLL10 UBE2J2 TNFRSF18 MIR200A MIR200B B3GALT6 SDF4 C1orf159 TNFRSF4 LOC105378948 TTLL10-AS1 MIR429 RNF223
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss SCNN1D FAM132A LINC01342 TTLL10 MIR6726 UBE2J2 TNFRSF18 MIR200A MIR200B B3GALT6 SDF4 C1orf159 TNFRSF4 TTLL10-AS1 MIR429 RNF223 ACAP3
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain SCNN1D FAM132A TTLL10 MIR6726 UBE2J2 TNFRSF18 PUSL1 MIR200A MIR200B B3GALT6 CPSF3L SDF4 MIR6727 TNFRSF4 TTLL10-AS1 MIR429 ACAP3

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline