This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01681381_cn
See other EIF4G3 CNV Assays ›
Gene Symbol
EIF4G3
Assay Reference Genome
Location

Chr.1:20817480 on build GRCh38
Cytoband
1p36.12
Assay ID Hs01681381_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8672

Gene Name:

eukaryotic translation initiation factor 4 gamma 3

Gene Aliases:

eIF-4G 3, eIF4G 3, eIF4GII

Location:

Chr.1:20806292-21177320 on Build GRCh38

Assay Gene Location:

Within Exon 40
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4G3 NM_001198801.1 32 4989 NP_001185730.1
NM_001198802.1 32 4899 NP_001185731.1
NM_003760.4 29 4515 NP_003751.2
XM_011542356.2 35 5589 XP_011540658.1
XM_011542361.2 34 5478 XP_011540663.1
XM_011542370.1 35 5592 XP_011540672.1
XM_011542371.1 33 5334 XP_011540673.1
XM_011542374.1 32 5301 XP_011540676.1
XM_011542375.2 32 6889 XP_011540677.1
XM_011542381.2 22 2975 XP_011540683.1
XM_017002672.1 36 7127 XP_016858161.1
XM_017002673.1 34 5571 XP_016858162.1
XM_017002674.1 34 5568 XP_016858163.1
XM_017002675.1 34 5559 XP_016858164.1
XM_017002676.1 33 5538 XP_016858165.1
XM_017002677.1 33 5415 XP_016858166.1
XM_017002678.1 34 5481 XP_016858167.1
XM_017002679.1 34 5478 XP_016858168.1
XM_017002680.1 31 4508 XP_016858169.1
XM_017002681.1 34 5475 XP_016858170.1
XM_017002682.1 33 5457 XP_016858171.1
XM_017002683.1 33 5457 XP_016858172.1
XM_017002684.1 34 6983 XP_016858173.1
XM_017002685.1 33 5448 XP_016858174.1
XM_017002686.1 33 5445 XP_016858175.1
XM_017002687.1 33 5445 XP_016858176.1
XM_017002688.1 30 4474 XP_016858177.1
XM_017002689.1 35 5592 XP_016858178.1
XM_017002690.1 33 5442 XP_016858179.1
XM_017002691.1 32 5427 XP_016858180.1
XM_017002692.1 32 5424 XP_016858181.1
XM_017002693.1 31 5295 XP_016858182.1
XM_017002694.1 33 5367 XP_016858183.1
XM_017002695.1 33 5364 XP_016858184.1
XM_017002696.1 32 5346 XP_016858185.1
XM_017002697.1 32 5343 XP_016858186.1
XM_017002698.1 32 5334 XP_016858187.1
XM_017002699.1 34 5481 XP_016858188.1
XM_017002700.1 31 5313 XP_016858189.1
XM_017002701.1 31 5310 XP_016858190.1
XM_017002702.1 31 5244 XP_016858191.1
XM_017002703.1 29 4405 XP_016858192.1
XM_017002704.1 33 5457 XP_016858193.1
XM_017002705.1 34 4752 XP_016858194.1
XM_017002706.1 33 4641 XP_016858195.1
XM_017002707.1 31 4584 XP_016858196.1
XM_017002708.1 30 4473 XP_016858197.1
XM_017002709.1 32 5336 XP_016858198.1
AB209119.1 32 4858 BAD92356.1
AF012072.2 29 4515 AAC02903.2
AK294883.1 23 3762
AK302873.1 20 2756
BC094683.1 27 3522 AAH94683.1
BC136643.1 29 4438
Z34918.1 13 1601 CAA84397.1

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-4gzsq:80/100.66.74.9:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0