Assay Details
Target Gene Details
Entrez Gene ID: | 740 |
Gene Name: | mitochondrial ribosomal protein L49 |
Gene Aliases: |
C11orf4, L49mt, MRP-L49, NOF, NOF1 |
Location: |
Chr.11:65122183-65127369 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRPL49 | NM_004927.3 | 4 | 553 | NP_004918.1 |
| NR_037567.1 | 4 | 530 | ||
| NR_037568.1 | 3 | 402 | ||
| AB062395.1 | 4 | 457 | BAB93482.1 | |
| AB209160.1 | 2 | 1947 | BAD92397.1 | |
| AK123252.1 | 2 | 984 | ||
| AK129795.1 | 4 | 474 | ||
| AK129986.1 | 4 | 478 | ||
| AK311821.1 | 4 | 476 | ||
| BC004378.1 | 4 | 461 | AAH04378.1 | |
| BG250495.1 | 4 | 541 | ||
| DA158218.1 | ||||
| DA162786.1 | 3 | 396 | ||
| DA484303.1 | 4 | 467 | ||
| U39400.1 | 4 | 463 | AAB18826.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526181 | Chr.11:64870569 - 65489858 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map