This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01689157_cn
See other SALL3 CNV Assays ›
Gene Symbol
SALL3
Assay Reference Genome
Location

Chr.18:78994576 on build GRCh38
Cytoband
18q23
Assay ID Hs01689157_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

27164

Gene Name:

spalt like transcription factor 3

Gene Aliases:

ZNF796

Location:

Chr.18:78980275-78998969 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SALL3 NM_171999.3 2 2585 NP_741996.2
BC148296.2 3 1778

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv577793 Chr.18:78951024 - 79022312 on Build GRCh38 Loss SALL3
nsv828333 Chr.18:78955949 - 79017302 on Build GRCh38 Loss SALL3
nsv577794 Chr.18:78951366 - 79472709 on Build GRCh38 Gain LOC107985149 LOC100996274 ATP9B SALL3 NFATC1
nsv577795 Chr.18:78972221 - 79050030 on Build GRCh38 Loss SALL3
nsv2377 Chr.18:78991267 - 79025451 on Build GRCh38 Insertion SALL3
dgv564n67 Chr.18:78952775 - 79013936 on Build GRCh38 Gain SALL3
nsv1065902 Chr.18:78538045 - 80257174 on Build GRCh38 Gain ADNP2 LOC107985149 KCNG2 LOC284241 LOC100996274 ATP9B LOC105376875 CTDP1 PQLC1 LOC284240 TXNL4A HSBP1L1 RBFA PARD6G SALL3 NFATC1 PARD6G-AS1 RBFADN
nsv828338 Chr.18:78963814 - 79001729 on Build GRCh38 Gain SALL3
esv2717497 Chr.18:78170182 - 79635788 on Build GRCh38 Deletion LOC107985149 LOC100996274 ATP9B SALL3 NFATC1 LOC284240
nsv1141945 Chr.18:78170394 - 79635820 on Build GRCh38 Deletion LOC107985149 LOC100996274 ATP9B SALL3 NFATC1 LOC284240

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
signal transduction
olfactory bulb interneuron development
neurogenesis
forelimb morphogenesis
hindlimb morphogenesis
negative regulation of smoothened signaling pathway
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-f2p7g:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline