This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01716692_cn
See other DHDDS CNV Assays ›
Gene Symbol
DHDDS
Assay Reference Genome
Location

Chr.1:26442819 on build GRCh38
Cytoband
1p36.11
Assay ID Hs01716692_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79947

Gene Name:

dehydrodolichyl diphosphate synthase subunit

Gene Aliases:

CIT, CPT, DS, HDS, RP59

Location:

Chr.1:26432282-26471306 on Build GRCh38

Assay Gene Location:

Within Exon 4
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DHDDS NM_001243564.1 4 419 NP_001230493.1
NM_001243565.1 4 419 NP_001230494.1
NM_001319959.1 4 442 NP_001306888.1
NM_024887.3 4 419 NP_079163.2
NM_205861.2 4 419 NP_995583.1
XM_006710912.2 4 478 XP_006710975.1
XM_006710913.2 4 357 XP_006710976.1
XM_006710914.2 4 354 XP_006710977.1
XM_006710915.2 4 391 XP_006710978.1
XM_006710916.2 4 468 XP_006710979.1
XM_006710917.2 4 421 XP_006710980.1
XM_006710918.2 4 377 XP_006710981.1
XM_011542183.2 4 454 XP_011540485.1
XM_011542184.2 4 475 XP_011540486.1
XM_011542185.2 4 475 XP_011540487.1
XM_011542186.2 4 475 XP_011540488.1
XM_017002379.1 4 357 XP_016857868.1
XM_017002380.1 4 454 XP_016857869.1
XM_017002381.1 4 468 XP_016857870.1
XM_017002382.1 4 377 XP_016857871.1
XM_017002383.1 4 468 XP_016857872.1
XM_017002384.1 4 421 XP_016857873.1
AB090852.1 3 280 BAC57588.1
AK023164.1 4 349 BAB14439.1
AK297134.1 3 339
AK308187.1 4 361
AK316485.1 4 362
BC003643.2 3 324 AAH03643.1
BC004117.2 4 361 AAH04117.1
BX648507.1 4 444
DA756178.1 4 422
GU727641.1 3 324

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
dolichyl diphosphate biosynthetic process
polyprenyltransferase activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0