This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01722103_cn
See other SLC2A4RG CNV Assays ›
Gene Symbol
SLC2A4RG
Assay Reference Genome
Location

Chr.20:63743473 on build GRCh38
Cytoband
20q13.33
Assay ID Hs01722103_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

56731

Gene Name:

SLC2A4 regulator

Gene Aliases:

GEF, HDBP-1, HDBP1, Si-1-2, Si-1-2-19

Location:

Chr.20:63739858-63744050 on Build GRCh38

Assay Gene Location:

Within Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
SLC2A4RG NM_020062.3 8 1702 NP_064446.2
AB044786.1 BAD29732.1
AB052777.1 BAD29733.1
AF249267.3 AAF97516.2
AV659783.2 1 38
BC001402.2 AAH01402.2
BC017446.1 AAH17446.1
BC028349.2 AAH28349.2
BC052306.1 AAH52306.1
BM983611.1 1 595

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion ZBTB46 ZGPAT LIME1 SLC2A4RG
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion ZBTB46-AS1 TNFRSF6B STMN3 ZBTB46 RTEL1-TNFRSF6B ZGPAT RTEL1 ARFRP1 LIME1 SLC2A4RG
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion TNFRSF6B STMN3 ZBTB46 RTEL1-TNFRSF6B ZGPAT RTEL1 ARFRP1 LIME1 SLC2A4RG
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss GMEB2 STMN3 UCKL1-AS1 MIR941-4 LINC00176 RTEL1-TNFRSF6B LINC00266-1 ZNF512B OPRL1 SLC2A4RG MIR6813 MIR941-1 MIR941-2 C20orf181 TCEA2 PCMTD2 MYT1 LIME1 MIR941-3 PRPF6 TNFRSF6B MIR941-5 TPD52L2 ZGPAT SOX18 MIR647 MIR1914 ZBTB46-AS1 UCKL1 NPBWR2 ZBTB46 LOC100505771 LKAAEAR1 RGS19 RTEL1 ABHD16B DNAJC5 SAMD10 ARFRP1
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss ZBTB46 ZGPAT LIME1 SLC2A4RG
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain ZBTB46 ZGPAT LIME1 SLC2A4RG
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss TNFRSF6B GMEB2 STMN3 RTEL1-TNFRSF6B ZGPAT EEF1A2 SRMS SLC2A4RG HELZ2 PTK6 ZBTB46 LOC100505771 RTEL1 ARFRP1 LIME1 FNDC11 PPDPF
dgv7684n54 Chr.20:63738644 - 63789659 on Build GRCh38 Loss ZBTB46 LIME1 SLC2A4RG
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss TNFRSF6B GMEB2 STMN3 RTEL1-TNFRSF6B ZGPAT KCNQ2 EEF1A2 SRMS SLC2A4RG HELZ2 PTK6 ZBTB46 LOC100505771 RTEL1 ARFRP1 LIME1 FNDC11 PPDPF

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-5xwv5:80/100.66.76.242:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0