This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01867063_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1254422 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01867063_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 7 1750 NP_477515.2
NM_194315.1 8 1798 NP_919296.1
NM_194457.1 6 1619 NP_919439.1
NM_194458.1 7 1879 NP_919440.1
XM_005244718.3 7 2028 XP_005244775.1
XM_005244719.4 7 1916 XP_005244776.1
XM_006710333.3 7 2195 XP_006710396.1
XM_011540613.2 6 1623 XP_011538915.1
XM_011540614.2 7 1981 XP_011538916.1
XM_011540616.2 8 1990 XP_011538918.1
XM_011540617.2 7 1757 XP_011538919.1
XM_017000238.1 8 1861 XP_016855727.1
XM_017000239.1 6 2079 XP_016855728.1
AF116687.2 1 114 AAF71107.1
AK056065.1 7 1660 BAB71086.1
AK098803.1 2 1116
BC094777.1 7 1749 AAH94777.1
BE515187.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication ACAP3 TNFRSF4 MIR200B CPSF3L CPTP TNFRSF18 UBE2J2 MIR200A LOC105378948 MIR6727 FAM132A LOC100288175 TTLL10 AGRN PUSL1 RNF223 B3GALT6 TTLL10-AS1 SDF4 TAS1R3 MIR6726 C1orf159 MIR429 SCNN1D LINC01342
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC148413 VWA1 NADK TNFRSF4 SSU72 TNFRSF18 UBE2J2 LINC00115 SLC35E2B DVL1 LOC102724312 LOC105378948 TMEM240 MIR6727 FAM132A LINC01128 TMEM88B MIR6808 CCNL2 PERM1 SAMD11 MMP23B TTLL10 MMP23A MRPL20 ATAD3C MXRA8 AURKAIP1 B3GALT6 TTLL10-AS1 LOC284600 SDF4 TAS1R3 MIR429 SCNN1D ATAD3A ACAP3 LOC100287934 LOC107985728 HES4 ISG15 NOC2L PLEKHN1 ANKRD65 LOC100130417 MIR200B CPSF3L CPTP LOC107985729 MIR200A FNDC10 LOC100288069 LOC100288175 AGRN CDK11A PUSL1 CDK11B SLC35E2 MIB2 FAM41C RNF223 FAM87B MIR6726 C1orf159 KLHL17 LINC01342 ATAD3B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss VWA1 OR4F29 TNFRSF4 TNFRSF18 UBE2J2 LINC00115 SLC35E2B LOC102724312 PRKCZ TMEM240 LOC100133331 FAM132A LINC01128 LOC107984841 GNB1 MMP23B TTLL10 MORN1 MRPL20 LOC100506504 AURKAIP1 B3GALT6 TTLL10-AS1 SDF4 TAS1R3 MIR429 SCNN1D ATAD3A FAM138A LOC107985728 ISG15 PLEKHN1 ANKRD65 CPTP LOC729737 WASH7P FNDC10 MTND1P23 AGRN CDK11B FAM41C TMEM52 FAM87B C1orf159 KLHL17 ATAD3B LOC148413 OR4F5 LOC105378589 NADK SSU72 DVL1 GABRD LOC105378948 LOC105378949 SKI MIR6727 TMEM88B MIR6808 CCNL2 PERM1 SAMD11 MMP23A LOC100134822 ATAD3C MXRA8 LOC100132287 LOC284600 CFAP74 LOC105378592 ACAP3 LOC100287934 MIR6723 HES4 LOC101928626 MIR6859-2 NOC2L LOC105378591 DDX11L1 LOC100130417 MIR200B CPSF3L FAAP20 MTND2P28 LOC105378947 LOC107985729 MIR200A LOC102725121 LOC100288069 OR4F16 LOC100288175 LOC100129534 CDK11A PUSL1 MIR6859-1 MIR1302-2 SLC35E2 MIB2 RNF223 CALML6 MIR6726 LINC01342
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss FAM132A UBE2J2 ACAP3 B3GALT6 SDF4 MIR6726 SCNN1D
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LOC107985728 HES4 ISG15 NOC2L PLEKHN1 TNFRSF4 LOC100130417 MIR200B TNFRSF18 UBE2J2 LINC00115 MIR200A LOC105378948 FAM132A LOC100288175 LINC01128 PERM1 SAMD11 TTLL10 AGRN FAM41C RNF223 B3GALT6 TTLL10-AS1 LOC284600 SDF4 FAM87B C1orf159 KLHL17 MIR429 LINC01342
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion ATAD3A LOC148413 ACAP3 VWA1 ANKRD65 TNFRSF4 SSU72 MIR200B CPSF3L CPTP TNFRSF18 UBE2J2 LOC107985729 DVL1 MIR200A LOC102724312 FNDC10 TMEM240 MIR6727 FAM132A TMEM88B MIR6808 CCNL2 MMP23B TTLL10 MRPL20 PUSL1 CDK11B ATAD3C MXRA8 AURKAIP1 MIB2 B3GALT6 TTLL10-AS1 SDF4 TAS1R3 MIR6726 C1orf159 MIR429 SCNN1D LINC01342 ATAD3B
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC148413 ACAP3 LOC107985728 HES4 ISG15 NOC2L PLEKHN1 ANKRD65 TNFRSF4 LOC100130417 MIR200B CPSF3L CPTP TNFRSF18 UBE2J2 DVL1 MIR200A LOC105378948 MIR6727 FAM132A LOC100288175 TMEM88B MIR6808 CCNL2 PERM1 SAMD11 TTLL10 MRPL20 AGRN PUSL1 MXRA8 AURKAIP1 RNF223 B3GALT6 TTLL10-AS1 LOC284600 SDF4 TAS1R3 MIR6726 C1orf159 KLHL17 MIR429 SCNN1D LINC01342
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain FAM132A TTLL10 TNFRSF4 MIR200B TNFRSF18 UBE2J2 RNF223 B3GALT6 TTLL10-AS1 SDF4 MIR200A LOC105378948 C1orf159 MIR429 SCNN1D LINC01342
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss FAM132A ACAP3 TTLL10 TNFRSF4 MIR200B TNFRSF18 UBE2J2 RNF223 B3GALT6 TTLL10-AS1 SDF4 MIR200A MIR6726 C1orf159 MIR429 SCNN1D LINC01342
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain FAM132A ACAP3 TTLL10 TNFRSF4 PUSL1 MIR200B CPSF3L TNFRSF18 UBE2J2 B3GALT6 TTLL10-AS1 SDF4 MIR200A MIR6726 MIR429 SCNN1D MIR6727

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline