Assay Details
Target Gene Details
Entrez Gene ID: | 7075 |
Gene Name: | tyrosine kinase with immunoglobulin like and EGF like domains 1 |
Gene Aliases: |
JTK14, TIE |
Location: |
Chr.1:43300895-43323110 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TIE1 | NM_001253357.1 | 13 | 2193 | NP_001240286.1 |
| NM_005424.4 | 13 | 2276 | NP_005415.1 | |
| XM_005271163.1 | 12 | 2090 | XP_005271220.1 | |
| XM_006710869.1 | 13 | 2410 | XP_006710932.1 | |
| XM_017002207.1 | 13 | 2219 | XP_016857696.1 | |
| AF217976.1 | 3 | 360 | AAG17219.1 | |
| AK225560.1 | 13 | 1935 | ||
| AK296735.1 | 8 | 1252 | ||
| AK300399.1 | 13 | 2083 | ||
| AK300623.1 | 13 | 1976 | ||
| AK307639.1 | 6 | 944 | ||
| AL833389.1 | ||||
| BC038239.1 | 13 | 2178 | AAH38239.1 | |
| EU826590.1 | 13 | 2099 | ||
| X60957.1 | 13 | 2135 | CAA43290.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546128 | Chr.1:43301467 - 43451738 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map