This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01888252_cn
See other MIB2 CNV Assays ›
Gene Symbol
MIB2
Assay Reference Genome
Location

Chr.1:1628136 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01888252_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

142678

Gene Name:

mindbomb E3 ubiquitin protein ligase 2

Gene Aliases:

ZZANK1, ZZZ5

Location:

Chr.1:1615188-1630610 on Build GRCh38

Assay Gene Location:

Overlaps Exon 15 - Intron 15
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIB2 NM_001170686.1 NP_001164157.1
NM_001170687.1 NP_001164158.1
NM_001170688.1 NP_001164159.1
NM_001170689.1 NP_001164160.1
NM_080875.2 NP_543151.2
NR_033183.1
XM_006710372.1 XP_006710435.1
XM_011540731.2 XP_011539033.1
XM_011540736.2 XP_011539038.1
XM_011540737.2 XP_011539039.1
XM_011540741.2 XP_011539043.1
XM_011540742.2 XP_011539044.1
XM_017000349.1 XP_016855838.1
XM_017000350.1 XP_016855839.1
XM_017000351.1 XP_016855840.1
XM_017000352.1 XP_016855841.1
XM_017000353.1 XP_016855842.1
XM_017000354.1 XP_016855843.1
XM_017000355.1 XP_016855844.1
XM_017000356.1 XP_016855845.1
XM_017000357.1 XP_016855846.1
XM_017000358.1 XP_016855847.1
XM_017000359.1 XP_016855848.1
XM_017000360.1 XP_016855849.1
XM_017000361.1 XP_016855850.1
XM_017000362.1 XP_016855851.1
XM_017000363.1 XP_016855852.1
XM_017000364.1 XP_016855853.1
XM_017000365.1 XP_016855854.1
XM_017000366.1 XP_016855855.1
AB074480.1 BAB92950.1
AB076691.1 BAC00992.1
AB076692.1 BAC00993.1
AB076693.1 BAC00994.1
AB097000.1 BAC77353.1
AK091610.1 BAC03707.1
AK095914.1 BAC04646.1
AK096295.1 BAC04752.1
AK097106.1 BAC04952.1
AK098785.1 BAC05413.1
AK122751.1
AK122863.1 14 2089
AK128167.1
AK128553.1
AK293642.1
AK302757.1
AK309362.1
AL834527.1 CAD39183.1
BC016490.1 AAH16490.1
BC037542.1 AAH37542.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MMP23A NADK SLC35E2 CDK11B CDK11A MIB2 MMP23B SLC35E2B GNB1 FNDC10
dgv8n100 Chr.1:1569349 - 1741152 on Build GRCh38 Loss MMP23A SLC35E2 CDK11B CDK11A MIB2 SSU72 MMP23B SLC35E2B FNDC10
esv3575582 Chr.1:1624086 - 1721336 on Build GRCh38 Gain MMP23A CDK11B CDK11A MIB2 MMP23B SLC35E2B
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss FAM87B SLC35E2 MRPL20 TTLL10-AS1 LOC102724312 TTLL10 SDF4 LINC00115 MMP23B TNFRSF18 LOC100288175 PERM1 CCNL2 B3GALT6 LOC284600 SSU72 PLEKHN1 ISG15 SLC35E2B LOC100288069 TAS1R3 CPTP CDK11B TMEM88B NADK RNF223 ANKRD65 MIR200A ACAP3 MIR6808 PUSL1 SAMD11 ATAD3B LOC107985729 MIR200B LOC148413 FAM41C DVL1 AURKAIP1 ATAD3A MIR429 AGRN UBE2J2 SCNN1D CDK11A FAM132A FNDC10 MMP23A MXRA8 CPSF3L MIR6726 KLHL17 LOC100130417 MIB2 LOC100287934 LINC01128 NOC2L MIR6727 LOC105378948 C1orf159 TNFRSF4 ATAD3C TMEM240 LOC107985728 HES4 VWA1 LINC01342
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC102724312 SDF4 MIR6859-2 MMP23B TNFRSF18 LOC100288175 LOC105378592 PERM1 SKI B3GALT6 OR4F5 LOC284600 PLEKHN1 ISG15 MTND1P23 LOC100288069 TAS1R3 GABRD CALML6 OR4F16 NADK ANKRD65 WASH7P ACAP3 LOC100134822 MIR6723 SAMD11 LOC105378947 LOC107985729 MIR200B LOC148413 FAM41C DVL1 AURKAIP1 ATAD3A MIR6859-1 FAM138A UBE2J2 LOC105378589 LOC100133331 FAM132A FNDC10 CFAP74 MIR6726 KLHL17 FAAP20 MIB2 LINC01128 OR4F29 LOC105378948 GNB1 ATAD3C LOC102725121 TMEM240 MORN1 HES4 VWA1 LOC101928626 FAM87B LOC100132287 SLC35E2 LOC100506504 MRPL20 TTLL10-AS1 TTLL10 PRKCZ LINC00115 TMEM52 DDX11L1 CCNL2 SSU72 SLC35E2B CPTP CDK11B TMEM88B LOC107984841 RNF223 MIR200A LOC729737 LOC105378591 MIR6808 PUSL1 ATAD3B MTND2P28 MIR429 AGRN SCNN1D CDK11A MMP23A MXRA8 CPSF3L LOC100130417 LOC100129534 LOC100287934 MIR1302-2 NOC2L MIR6727 C1orf159 TNFRSF4 LOC107985728 LINC01342 LOC105378949
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MMP23A NADK SLC35E2 CDK11B CDK11A MIB2 SSU72 MMP23B SLC35E2B GNB1 FNDC10
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion PUSL1 MRPL20 ATAD3B TTLL10-AS1 LOC102724312 TTLL10 LOC107985729 MIR200B LOC148413 SDF4 DVL1 AURKAIP1 MMP23B TNFRSF18 ATAD3A MIR429 CCNL2 UBE2J2 B3GALT6 SCNN1D SSU72 FAM132A FNDC10 MXRA8 CPSF3L TAS1R3 MIR6726 CPTP CDK11B MIB2 MIR6727 TMEM88B C1orf159 TNFRSF4 ATAD3C ANKRD65 TMEM240 VWA1 MIR200A LINC01342 ACAP3 MIR6808
esv2661270 Chr.1:1625365 - 1636030 on Build GRCh38 Deletion CDK11B MIB2 MMP23B
esv2760410 Chr.1:1565561 - 1756248 on Build GRCh38 Gain+Loss MMP23A NADK SLC35E2 CDK11B CDK11A MIB2 SSU72 MMP23B SLC35E2B FNDC10
nsv545017 Chr.1:1615322 - 1663759 on Build GRCh38 Loss CDK11B MIB2 MMP23B SLC35E2B
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss SLC35E2 MRPL20 ATAD3B LOC102724312 LOC107985729 LOC148413 PRKCZ TMEM52 MMP23B ATAD3A CCNL2 CDK11A SSU72 LOC105378589 SLC35E2B FNDC10 CFAP74 MMP23A GABRD CDK11B MIB2 CALML6 TMEM88B GNB1 NADK ATAD3C ANKRD65 TMEM240 VWA1 LOC105378591 LOC105378949

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
Notch signaling pathway
positive regulation of I-kappaB kinase/NF-kappaB signaling
actin binding
ubiquitin-protein transferase activity
signal transducer activity
protein binding
zinc ion binding
ligase activity
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline