This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01890904_cn
See other LIME1 CNV Assays ›
Gene Symbol
LIME1
Assay Reference Genome
Location

Chr.20:63735791 on build GRCh38
Cytoband
20q13.33
Assay ID Hs01890904_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54923

Gene Name:

Lck interacting transmembrane adaptor 1

Gene Aliases:

LIME, dJ583P15.4

Location:

Chr.20:63735701-63739107 on Build GRCh38

Assay Gene Location:

Within Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LIME1 NM_001305654.1 1 91 NP_001292583.1
AY203957.1 1 91 AAP34480.1

Target Gene Details

Entrez Gene ID:

84619

Gene Name:

zinc finger CCCH-type and G-patch domain containing

Gene Aliases:

GPATC6, GPATCH6, KIAA1847, ZC3H9, ZC3HDC9, ZIP

Location:

Chr.20:63707442-63736142 on Build GRCh38

Assay Gene Location:

Overlaps Intron 7 - Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ZGPAT NM_001083113.1 NP_001076582.1
NM_001195653.1 NP_001182582.1
NM_001195654.1 NP_001182583.1
NM_032527.4 NP_115916.3
NM_181485.2 NP_852150.2
AB058750.3
AK027878.1 BAB55426.1
AK074961.1 BAC11317.1
AK307368.1
AK314726.1
BC019338.1 AAH19338.1
BC032612.1 AAH32612.1
KU178806.1
KU178807.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion LIME1 ZGPAT SLC2A4RG ZBTB46
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion STMN3 LIME1 RTEL1-TNFRSF6B ZGPAT ZBTB46-AS1 ARFRP1 SLC2A4RG RTEL1 TNFRSF6B ZBTB46
nsv586614 Chr.20:63725275 - 63741460 on Build GRCh38 Gain LIME1 ZGPAT SLC2A4RG
nsv1139762 Chr.20:63731048 - 63737647 on Build GRCh38 Duplication LIME1 ZGPAT
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion STMN3 LIME1 RTEL1-TNFRSF6B ZGPAT ARFRP1 SLC2A4RG RTEL1 TNFRSF6B ZBTB46
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss RGS19 NPBWR2 SOX18 ARFRP1 PCMTD2 MIR941-4 SLC2A4RG TNFRSF6B ZNF512B LKAAEAR1 LINC00176 MIR6813 C20orf181 LIME1 MIR647 RTEL1-TNFRSF6B UCKL1 DNAJC5 RTEL1 UCKL1-AS1 LOC100505771 MIR941-2 MIR941-5 ABHD16B LINC00266-1 MIR1914 MIR941-3 OPRL1 ZGPAT PRPF6 GMEB2 MYT1 STMN3 TPD52L2 MIR941-1 ZBTB46-AS1 SAMD10 TCEA2 ZBTB46
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss LIME1 ZGPAT SLC2A4RG ZBTB46
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain LIME1 ZGPAT SLC2A4RG ZBTB46
nsv586615 Chr.20:63734204 - 63741603 on Build GRCh38 Gain LIME1 ZGPAT SLC2A4RG
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss ZGPAT HELZ2 GMEB2 ARFRP1 SLC2A4RG FNDC11 TNFRSF6B STMN3 LIME1 PPDPF RTEL1-TNFRSF6B SRMS RTEL1 LOC100505771 ZBTB46 EEF1A2 PTK6
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss KCNQ2 ZGPAT HELZ2 GMEB2 ARFRP1 SLC2A4RG FNDC11 TNFRSF6B STMN3 LIME1 PPDPF RTEL1-TNFRSF6B SRMS RTEL1 LOC100505771 ZBTB46 EEF1A2 PTK6

Back To Top

More Information


Set Membership:

Intragenic Exonic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
T cell receptor signaling pathway
B cell receptor signaling pathway
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of epidermal growth factor-activated receptor activity
negative regulation of transcription, DNA-templated
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
sequence-specific DNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline