This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01934309_cn
See other ADAM15 CNV Assays ›
Gene Symbol
ADAM15
Assay Reference Genome
Location

Chr.1:155054433 on build GRCh38
Cytoband
1q21.3
Assay ID Hs01934309_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,496.00
Your Price
Online offer:
4496.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8751

Gene Name:

ADAM metallopeptidase domain 15

Gene Aliases:

MDC15

Location:

Chr.1:155051272-155062776 on Build GRCh38

Assay Gene Location:

Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ADAM15 NM_001261464.1 6 684 NP_001248393.1
NM_001261465.1 6 654 NP_001248394.1
NM_001261466.1 6 606 NP_001248395.1
NM_003815.4 6 654 NP_003806.3
NM_207191.2 6 654 NP_997074.1
NM_207194.2 6 654 NP_997077.1
NM_207195.2 6 654 NP_997078.1
NM_207196.2 6 654 NP_997079.1
NM_207197.2 6 654 NP_997080.1
NR_048577.1 6 654
NR_048578.1 6 654
NR_048579.1 6 654
AB209157.1 5 2185 BAD92394.1
AK000667.1 6 640
AK075498.1
AK295614.1 6 626
AK296925.1 6 605
AK297468.1 6 527
AK308172.1 6 577
AY518542.1 6 539 AAR99331.1
AY560593.1 6 640 AAS72991.1
AY560594.1 6 640 AAS72992.1
AY560595.1 6 640 AAS72993.1
AY560596.1 6 640 AAS72994.1
AY560597.1 6 640 AAS72995.1
AY560598.1 6 640 AAS72996.1
AY560599.1 6 640 AAS72997.1
AY560600.1 6 640 AAS72998.1
AY560601.1 6 640 AAS72999.1
AY576417.1 6 640 AAS73000.1
BC014566.2 6 620 AAH14566.1
BT009764.1 6 539 AAP88766.1
U41767.1 6 546 AAC50404.1
U46005.1 6 539 AAC51112.1

Target Gene Details

Entrez Gene ID:

100505666

Gene Name:

uncharacterized LOC100505666

Gene Aliases:

-

Location:

Chr.1:155045191-155063991 on Build GRCh38

Assay Gene Location:

Within Intron 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LOC100505666 NR_040773.1
BM047608.1
DB152307.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv509502 Chr.1:154904407 - 155142291 on Build GRCh38 Insertion PBXIP1 FLAD1 EFNA4 EFNA3 LOC101928120 LOC100505666 LENEP ZBTB7B SLC50A1 DPM3 PMVK SHC1 EFNA1 CKS1B PYGO2 DCST2 ADAM15 DCST1 MIR4258
esv3587615 Chr.1:155047853 - 155175013 on Build GRCh38 Gain EFNA1 EFNA4 EFNA3 LOC100505666 ADAM15 TRIM46 DCST1 KRTCAP2 SLC50A1 DPM3
nsv952212 Chr.1:154998225 - 155087624 on Build GRCh38 Deletion EFNA4 EFNA3 LOC100505666 DCST2 ADAM15 DCST1 ZBTB7B

Back To Top

More Information


Set Membership:

Intragenic Exonic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
negative regulation of cell-matrix adhesion
proteolysis
apoptotic process
cell-matrix adhesion
integrin-mediated signaling pathway
male gonad development
extracellular matrix disassembly
negative regulation of cell growth
negative regulation of cell migration
collagen catabolic process
tissue regeneration
innate immune response
cardiac epithelial to mesenchymal transition
protein kinase C signaling
negative regulation of receptor binding
metalloendopeptidase activity
integrin binding
protein binding
metallopeptidase activity
zinc ion binding
SH3 domain binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-v4fgr:80/100.66.76.147:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline