This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01957119_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1255046 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01957119_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 7 1126 NP_477515.2
NM_194315.1 8 1174 NP_919296.1
NM_194457.1 6 995 NP_919439.1
NM_194458.1 7 1255 NP_919440.1
XM_005244718.3 7 1404 XP_005244775.1
XM_005244719.4 7 1292 XP_005244776.1
XM_006710333.3 7 1571 XP_006710396.1
XM_011540613.2 6 999 XP_011538915.1
XM_011540614.2 7 1357 XP_011538916.1
XM_011540616.2 8 1366 XP_011538918.1
XM_011540617.2 7 1133 XP_011538919.1
XM_017000238.1 8 1237 XP_016855727.1
XM_017000239.1 6 1455 XP_016855728.1
AF296658.1 7 1079 AAK52609.1
AK056065.1 7 1036 BAB71086.1
AK075017.1 7 1105 BAC11355.1
AK098803.1 2 492
AK291935.1 7 1105
AW411508.1
BC094777.1 7 1125 AAH94777.1
BC104890.1 6 950 AAI04891.1
BC113430.1 6 950
BC143657.1 6 950
BQ881730.1 5 815

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication LOC100288175 SCNN1D TNFRSF18 CPSF3L B3GALT6 MIR6727 FAM132A C1orf159 MIR429 SDF4 PUSL1 MIR200B MIR200A TTLL10 MIR6726 RNF223 TTLL10-AS1 LINC01342 AGRN TNFRSF4 LOC105378948 UBE2J2 ACAP3 CPTP TAS1R3
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss LOC100288175 MMP23A MMP23B SLC35E2B LOC107985728 NADK DVL1 CPSF3L LOC148413 FNDC10 NOC2L LOC102724312 SDF4 TMEM240 LINC00115 MXRA8 TTLL10 FAM87B RNF223 LOC107985729 AGRN PLEKHN1 LOC284600 TNFRSF4 LOC100287934 LOC105378948 SLC35E2 UBE2J2 ACAP3 AURKAIP1 MRPL20 FAM41C MIB2 ATAD3C VWA1 SAMD11 SCNN1D ATAD3B TNFRSF18 ATAD3A HES4 B3GALT6 CDK11A MIR6727 ISG15 FAM132A C1orf159 MIR429 KLHL17 LINC01128 PUSL1 LOC100288069 MIR6808 MIR200B MIR200A TMEM88B MIR6726 CCNL2 ANKRD65 TTLL10-AS1 LOC100130417 LINC01342 CDK11B PERM1 SSU72 CPTP TAS1R3
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss OR4F5 SLC35E2B LOC107985728 MIR6859-2 CPSF3L LOC148413 TMEM240 LOC101928626 LOC100133331 LOC107985729 MIR6723 LOC284600 TNFRSF4 UBE2J2 ACAP3 MRPL20 FAM41C LOC100506504 SCNN1D TMEM52 ATAD3A SKI HES4 B3GALT6 WASH7P LOC105378589 MORN1 LOC105378947 CDK11A ISG15 C1orf159 DDX11L1 PRKCZ LOC100288069 TMEM88B MIR6726 CCNL2 ANKRD65 TTLL10-AS1 MTND1P23 LOC100130417 LINC01342 CALML6 CDK11B MIR6859-1 CFAP74 GNB1 LOC100288175 MMP23A MMP23B NADK LOC105378591 FAM138A DVL1 MTND2P28 FNDC10 LOC100134822 NOC2L LOC102724312 SDF4 LINC00115 LOC105378949 MXRA8 TTLL10 FAM87B RNF223 AGRN PLEKHN1 LOC729737 LOC100287934 LOC105378948 SLC35E2 LOC100129534 FAAP20 AURKAIP1 LOC100132287 MIB2 ATAD3C VWA1 SAMD11 ATAD3B TNFRSF18 MIR6727 FAM132A MIR429 KLHL17 LINC01128 LOC102725121 PUSL1 MIR6808 MIR200B MIR200A GABRD LOC107984841 OR4F29 MIR1302-2 PERM1 SSU72 CPTP TAS1R3 OR4F16 LOC105378592
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss MIR6726 UBE2J2 ACAP3 SCNN1D SDF4 B3GALT6 FAM132A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss LOC100288175 SAMD11 LOC107985728 TNFRSF18 HES4 B3GALT6 NOC2L ISG15 FAM132A C1orf159 MIR429 KLHL17 LINC01128 SDF4 LINC00115 MIR200B MIR200A TTLL10 FAM87B RNF223 TTLL10-AS1 LOC100130417 LINC01342 AGRN PLEKHN1 LOC284600 TNFRSF4 LOC105378948 UBE2J2 FAM41C PERM1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MMP23B SCNN1D ATAD3B TNFRSF18 DVL1 CPSF3L ATAD3A LOC148413 FNDC10 B3GALT6 MIR6727 FAM132A C1orf159 MIR429 LOC102724312 SDF4 PUSL1 TMEM240 MIR6808 MIR200B MXRA8 MIR200A TMEM88B TTLL10 MIR6726 CCNL2 ANKRD65 TTLL10-AS1 LOC107985729 LINC01342 TNFRSF4 UBE2J2 ACAP3 CDK11B AURKAIP1 MRPL20 SSU72 CPTP TAS1R3 MIB2 ATAD3C VWA1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain LOC100288175 SAMD11 LOC107985728 SCNN1D TNFRSF18 DVL1 CPSF3L LOC148413 HES4 B3GALT6 NOC2L MIR6727 ISG15 FAM132A C1orf159 MIR429 KLHL17 SDF4 PUSL1 MIR6808 MIR200B MXRA8 MIR200A TMEM88B TTLL10 MIR6726 CCNL2 ANKRD65 RNF223 TTLL10-AS1 LOC100130417 LINC01342 AGRN PLEKHN1 LOC284600 TNFRSF4 LOC105378948 UBE2J2 ACAP3 AURKAIP1 MRPL20 PERM1 CPTP TAS1R3
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain TTLL10 SCNN1D RNF223 TTLL10-AS1 TNFRSF18 B3GALT6 LINC01342 FAM132A TNFRSF4 C1orf159 MIR429 LOC105378948 UBE2J2 SDF4 MIR200B MIR200A
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss TTLL10 MIR6726 SCNN1D RNF223 TTLL10-AS1 TNFRSF18 B3GALT6 LINC01342 FAM132A TNFRSF4 C1orf159 MIR429 UBE2J2 ACAP3 SDF4 MIR200B MIR200A
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain TTLL10 MIR6726 SCNN1D TTLL10-AS1 TNFRSF18 CPSF3L B3GALT6 MIR6727 FAM132A TNFRSF4 MIR429 UBE2J2 ACAP3 SDF4 PUSL1 MIR200B MIR200A

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline