This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01974989_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1793333 on build GRCh38
Cytoband
1p36.33
Assay ID Hs01974989_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Overlaps Intron 8 - Exon 9
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 NP_001269467.1
NM_001282539.1 NP_001269468.1
NM_002074.4 NP_002065.1
XM_017001059.1 XP_016856548.1
XM_017001060.1 XP_016856549.1
XM_017001061.1 XP_016856550.1
AF070603.1 AAC28655.1
AF501882.1 AAM15918.1
AK096614.1 1 865
AK122949.1
AK312614.1
BC004186.2 AAH04186.1
BC005888.2 AAH05888.1
BC008991.1 AAH08991.1
BC114618.1 AAI14619.1
BT007305.1 AAP35969.1
CR456784.1 CAG33065.1
M36430.1 AAA63265.1
X04526.1 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion NADK MIB2 GNB1 MMP23B FNDC10 MMP23A CDK11A SLC35E2 CDK11B SLC35E2B
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss NADK GNB1 MMP23A CDK11A SLC35E2
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication NADK MIB2 GNB1 SSU72 MMP23B FNDC10 MMP23A CDK11A SLC35E2 CDK11B SLC35E2B
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC100506504 GABRD ACAP3 TNFRSF18 FAM87B LOC105378948 MIR6727 MIR200A ISG15 ANKRD65 LOC107985729 LOC100134822 AGRN AURKAIP1 SKI SLC35E2B NADK OR4F16 SDF4 SSU72 LOC105378589 ATAD3A LOC100288069 LOC100130417 DDX11L1 TMEM240 LOC105378591 SLC35E2 MTND1P23 PUSL1 MIR6808 MIR429 NOC2L CFAP74 LINC01128 C1orf159 KLHL17 CDK11A LOC100129534 CDK11B PRKCZ OR4F29 ATAD3B FNDC10 MIR6723 VWA1 DVL1 LOC105378947 LOC107985728 LOC105378949 CCNL2 RNF223 OR4F5 WASH7P MRPL20 MIR6859-2 SCNN1D TMEM88B MIR200B ATAD3C MIR6726 CPTP LOC100287934 LOC729737 LINC00115 LOC100288175 LINC01342 MIB2 B3GALT6 MORN1 SAMD11 MIR6859-1 TAS1R3 UBE2J2 LOC101928626 MMP23A CPSF3L LOC100132287 PERM1 TTLL10 LOC107984841 FAAP20 HES4 TMEM52 MMP23B PLEKHN1 GNB1 LOC148413 LOC102724312 FAM138A CALML6 TNFRSF4 MTND2P28 LOC102725121 FAM132A LOC100133331 MIR1302-2 TTLL10-AS1 MXRA8 FAM41C LOC105378592 LOC284600
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication NADK GNB1 LOC105378949
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss CCNL2 TMEM240 GABRD LOC105378591 MRPL20 SLC35E2 TMEM88B ATAD3C CFAP74 TMEM52 MMP23B CDK11A CDK11B PRKCZ MIB2 GNB1 ANKRD65 LOC148413 LOC107985729 LOC102724312 ATAD3B FNDC10 CALML6 SLC35E2B NADK VWA1 SSU72 LOC105378589 ATAD3A LOC105378949 MMP23A

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-bs9p2:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline