This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01976501_cn
See other MEGF6 CNV Assays ›
Gene Symbol
MEGF6
Assay Reference Genome
Location

Chr.1:3489458 on build GRCh38
Cytoband
1p36.32
Assay ID Hs01976501_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

1953

Gene Name:

multiple EGF like domains 6

Gene Aliases:

EGFL3

Location:

Chr.1:3487942-3624757 on Build GRCh38

Assay Gene Location:

Within Exon 41
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MEGF6 NM_001409.3 37 5923 NP_001400.3
XM_006710406.3 34 5558 XP_006710469.1
XM_011540885.1 38 6228 XP_011539187.1
XM_011540886.1 37 6099 XP_011539188.1
XM_011540887.2 33 5436 XP_011539189.1
XM_017000533.1 37 6096 XP_016856022.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2758915 Chr.1:3372374 - 3632289 on Build GRCh38 Gain MIR551A ARHGEF16 MEGF6 PRDM16 WRAP73 TPRG1L
nsv470687 Chr.1:3356499 - 3763210 on Build GRCh38 Loss MIR551A ARHGEF16 TP73-AS1 CCDC27 MEGF6 PRDM16 WRAP73 TP73 TPRG1L
nsv1012423 Chr.1:3477686 - 4747449 on Build GRCh38 Gain MIR551A TP73-AS1 CCDC27 MEGF6 WRAP73 CEP104 AJAP1 LINC01134 ARHGEF16 SMIM1 C1orf174 LRRC47 TP73 TPRG1L DFFB LOC284661 LINC01346
nsv951959 Chr.1:3051037 - 3812036 on Build GRCh38 Deletion MIR551A MIR4251 TP73-AS1 CCDC27 MEGF6 PRDM16 LOC105378604 WRAP73 ARHGEF16 SMIM1 LINC00982 LRRC47 TP73 TPRG1L
esv3893545 Chr.1:3460878 - 3512044 on Build GRCh38 Loss ARHGEF16 MEGF6
nsv830203 Chr.1:3446374 - 3607752 on Build GRCh38 Loss MIR551A ARHGEF16 MEGF6
nsv299 Chr.1:3476928 - 3512705 on Build GRCh38 Insertion ARHGEF16 MEGF6

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline