Assay Details
Target Gene Details
Entrez Gene ID: | 7088 |
Gene Name: | transducin like enhancer of split 1 |
Gene Aliases: |
ESG, ESG1, GRG1 |
Location: |
Chr.9:81583683-81689535 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TLE1 | NM_001303103.1 | 20 | 3894 | NP_001290032.1 |
| NM_001303104.1 | 20 | 3819 | NP_001290033.1 | |
| NM_005077.4 | 20 | 3864 | NP_005068.2 | |
| XM_005252151.1 | 21 | 2910 | XP_005252208.1 | |
| XM_005252152.1 | 21 | 2907 | XP_005252209.1 | |
| XM_005252153.1 | 21 | 2880 | XP_005252210.1 | |
| XM_005252154.1 | 21 | 2877 | XP_005252211.1 | |
| XM_005252162.1 | 19 | 2658 | XP_005252219.1 | |
| XM_005252163.2 | 18 | 2556 | XP_005252220.1 | |
| XM_006717258.1 | 21 | 2907 | XP_006717321.1 | |
| XM_006717259.3 | 19 | 2593 | XP_006717322.1 | |
| XM_006717260.1 | 19 | 2688 | XP_006717323.1 | |
| XM_006717261.2 | 19 | 2622 | XP_006717324.1 | |
| XM_006717262.1 | 20 | 2775 | XP_006717325.1 | |
| XM_006717263.1 | 20 | 2772 | XP_006717326.1 | |
| XM_011518951.2 | 20 | 2775 | XP_011517253.1 | |
| XM_017015064.1 | 20 | 2805 | XP_016870553.1 | |
| XM_017015065.1 | 20 | 2802 | XP_016870554.1 | |
| XM_017015066.1 | 18 | 2548 | XP_016870555.1 | |
| AB209854.1 | 18 | 6993 | BAD93091.1 | |
| BC010100.2 | 20 | 3010 | AAH10100.1 | |
| BC015747.1 | 20 | 2720 | AAH15747.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv821665 | Chr.9:80752718 - 82292739 on Build GRCh38 | Loss |
|
| nsv831638 | Chr.9:81538144 - 81707769 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map