This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02033887_cn
See other CPSF3L CNV Assays ›
Gene Symbol
CPSF3L
Assay Reference Genome
Location

Chr.1:1314297 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02033887_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54973

Gene Name:

cleavage and polyadenylation specific factor 3-like

Gene Aliases:

CPSF73L, INT11, INTS11, RC-68, RC68

Location:

Chr.1:1311585-1324687 on Build GRCh38

Assay Gene Location:

Overlaps Intron 10 - Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CPSF3L NM_001256456.1 NP_001243385.1
NM_001256460.1 NP_001243389.1
NM_001256462.1 NP_001243391.1
NM_001256463.1 NP_001243392.1
NM_017871.5 NP_060341.2
XM_011541647.1 XP_011539949.1
XM_011541648.1 XP_011539950.1
XM_011541650.1 XP_011539952.1
XM_017001557.1 XP_016857046.1
XM_017001558.1 XP_016857047.1
AK021939.1 BAB13943.1
AK023356.1 BAB14541.1
AK056652.1
AK058018.1
AK092190.1
AK123641.1 1 874
AK291387.1
AK297350.1
AL136813.1 CAB66747.1
BC000675.2 AAH00675.2
BC007978.2 AAH07978.1
BC008041.2 AAH08041.1
BC013904.2 AAH13904.2
BC020199.1 AAH20199.1
CR533557.1 CAG38588.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss TAS1R3 MIR200A NOC2L ATAD3A KLHL17 TMEM240 LOC729737 ATAD3C ISG15 LOC105378592 LOC105378589 ANKRD65 LOC105378591 TTLL10-AS1 LOC148413 PERM1 LOC107985728 CDK11A B3GALT6 TMEM88B MTND2P28 UBE2J2 MIR200B MIB2 FAM41C PRKCZ DDX11L1 SCNN1D MIR6859-1 CALML6 FNDC10 MIR6808 MMP23B NADK MIR429 CPTP MTND1P23 FAM138A LOC105378949 C1orf159 SLC35E2 LOC102725121 LOC284600 MMP23A VWA1 MIR6723 CDK11B LINC01342 LINC00115 PLEKHN1 MORN1 HES4 MIR6726 OR4F16 LOC100133331 LOC100287934 LOC100288175 LOC105378948 SAMD11 LOC100134822 LOC100288069 SKI LOC107985729 LOC107984841 TMEM52 AURKAIP1 FAAP20 MXRA8 LOC102724312 DVL1 LOC101928626 AGRN LOC100506504 ATAD3B CFAP74 MRPL20 OR4F5 OR4F29 WASH7P LOC100130417 LINC01128 FAM132A TNFRSF18 ACAP3 SSU72 TTLL10 TNFRSF4 RNF223 GNB1 MIR1302-2 MIR6859-2 LOC100129534 MIR6727 PUSL1 GABRD CPSF3L SDF4 SLC35E2B LOC105378947 LOC100132287 CCNL2 FAM87B
nsv544971 Chr.1:1294063 - 1351426 on Build GRCh38 Loss MIR6726 CPTP TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 MIR6727 PUSL1
nsv544969 Chr.1:1290683 - 1374597 on Build GRCh38 Loss MIR6726 CPTP MXRA8 TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 SCNN1D MIR6727 PUSL1 AURKAIP1
nsv997291 Chr.1:1306835 - 1376336 on Build GRCh38 Gain CPTP MXRA8 TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 MIR6727 PUSL1 AURKAIP1
nsv470680 Chr.1:1070426 - 1378635 on Build GRCh38 Loss MIR6726 TAS1R3 MIR6808 MIR200A MIR429 CPTP FAM132A TNFRSF18 ACAP3 TTLL10 C1orf159 TTLL10-AS1 TNFRSF4 RNF223 B3GALT6 MIR6727 PUSL1 LINC01342 AURKAIP1 UBE2J2 MXRA8 MIR200B DVL1 CPSF3L SDF4 SCNN1D
dgv6n100 Chr.1:1301750 - 1362354 on Build GRCh38 Gain+Loss CPTP MXRA8 TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 MIR6727 PUSL1
nsv1160788 Chr.1:1282706 - 1376336 on Build GRCh38 Deletion MIR6726 CPTP MXRA8 TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 SCNN1D MIR6727 PUSL1 AURKAIP1
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication MIR6726 TAS1R3 MIR200A MIR429 CPTP LOC100288175 FAM132A TNFRSF18 LOC105378948 ACAP3 TTLL10 C1orf159 TTLL10-AS1 TNFRSF4 RNF223 B3GALT6 MIR6727 PUSL1 LINC01342 UBE2J2 MIR200B CPSF3L SDF4 SCNN1D AGRN
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss MIR6726 TAS1R3 MIR200A NOC2L ATAD3A LOC100287934 LOC100288175 KLHL17 LOC105378948 TMEM240 ATAD3C SAMD11 ISG15 LOC100288069 ANKRD65 LOC107985729 TTLL10-AS1 LOC148413 PERM1 LOC107985728 CDK11A B3GALT6 TMEM88B AURKAIP1 UBE2J2 MXRA8 LOC102724312 MIR200B DVL1 MIB2 FAM41C SCNN1D AGRN ATAD3B FNDC10 MRPL20 MIR6808 MMP23B NADK LOC100130417 MIR429 CPTP LINC01128 FAM132A TNFRSF18 ACAP3 SSU72 TTLL10 C1orf159 SLC35E2 TNFRSF4 LOC284600 MMP23A RNF223 VWA1 CDK11B MIR6727 PUSL1 LINC01342 LINC00115 PLEKHN1 CPSF3L SDF4 SLC35E2B HES4 CCNL2 FAM87B
esv3890636 Chr.1:1304555 - 1353203 on Build GRCh38 Loss CPTP MXRA8 TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 MIR6727 PUSL1
nsv1000169 Chr.1:1281678 - 1362354 on Build GRCh38 Gain MIR6726 CPTP MXRA8 TAS1R3 DVL1 MIR6808 CPSF3L ACAP3 SCNN1D MIR6727 PUSL1
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion MIR6726 TAS1R3 FNDC10 MRPL20 MIR6808 MIR200A MMP23B ATAD3A MIR429 CPTP FAM132A TNFRSF18 TMEM240 ATAD3C ACAP3 SSU72 TTLL10 ANKRD65 C1orf159 LOC107985729 TTLL10-AS1 LOC148413 TNFRSF4 B3GALT6 VWA1 CDK11B MIR6727 TMEM88B PUSL1 LINC01342 AURKAIP1 UBE2J2 MXRA8 LOC102724312 MIR200B DVL1 CPSF3L MIB2 SDF4 SCNN1D CCNL2 ATAD3B
esv2758912 Chr.1:1263940 - 1384492 on Build GRCh38 Loss MIR6726 TAS1R3 MIR6808 MIR6727 PUSL1 AURKAIP1 UBE2J2 CPTP MXRA8 DVL1 CPSF3L ACAP3 SCNN1D
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain MIR6726 TAS1R3 MRPL20 MIR6808 MIR200A NOC2L LOC100130417 MIR429 CPTP LOC100288175 KLHL17 FAM132A TNFRSF18 LOC105378948 SAMD11 ISG15 ACAP3 TTLL10 ANKRD65 C1orf159 TTLL10-AS1 LOC148413 TNFRSF4 LOC284600 PERM1 RNF223 LOC107985728 B3GALT6 MIR6727 TMEM88B PUSL1 LINC01342 AURKAIP1 UBE2J2 MXRA8 PLEKHN1 MIR200B DVL1 CPSF3L SDF4 SCNN1D AGRN HES4 CCNL2
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR6726 TTLL10-AS1 TNFRSF4 B3GALT6 MIR200A MIR6727 PUSL1 MIR429 UBE2J2 FAM132A TNFRSF18 MIR200B CPSF3L ACAP3 SDF4 SCNN1D TTLL10

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

RNA processing factor

Gene Ontology Categories:

Function(s) Process(es)

snRNA processing
snRNA transcription from RNA polymerase II promoter
protein binding
hydrolase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0