This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02086596_cn
See other LIME1 CNV Assays ›
Gene Symbol
LIME1
Assay Reference Genome
Location

Chr.20:63736666 on build GRCh38
Cytoband
20q13.33
Assay ID Hs02086596_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54923

Gene Name:

Lck interacting transmembrane adaptor 1

Gene Aliases:

LIME, dJ583P15.4

Location:

Chr.20:63735701-63739107 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
LIME1 NM_001305654.1 NP_001292583.1
NM_001305655.1 1 207 NP_001292584.1
NM_017806.3 1 207 NP_060276.2
AK000413.1 BAA91148.1
AK054722.1 1 349
AY203957.1 AAP34480.1
BC017016.1 AAH17016.1
BI911934.1 1 233
BP368475.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2722959 Chr.20:63715664 - 63806918 on Build GRCh38 Deletion SLC2A4RG ZGPAT LIME1 ZBTB46
nsv953308 Chr.20:63650148 - 63831847 on Build GRCh38 Deletion RTEL1-TNFRSF6B TNFRSF6B ZBTB46-AS1 SLC2A4RG ZGPAT LIME1 ZBTB46 STMN3 RTEL1 ARFRP1
nsv586614 Chr.20:63725275 - 63741460 on Build GRCh38 Gain SLC2A4RG ZGPAT LIME1
nsv1139762 Chr.20:63731048 - 63737647 on Build GRCh38 Duplication ZGPAT LIME1
nsv1160690 Chr.20:63639552 - 63744287 on Build GRCh38 Deletion RTEL1-TNFRSF6B TNFRSF6B SLC2A4RG ZGPAT LIME1 ZBTB46 STMN3 RTEL1 ARFRP1
nsv586618 Chr.20:63736025 - 63740688 on Build GRCh38 Gain SLC2A4RG ZGPAT LIME1
nsv428381 Chr.20:63583375 - 64334167 on Build GRCh38 Gain+Loss MIR941-5 MIR941-2 ZBTB46-AS1 TPD52L2 SLC2A4RG ZGPAT LIME1 LOC100505771 NPBWR2 MYT1 TNFRSF6B LINC00176 MIR647 MIR941-4 SOX18 LKAAEAR1 MIR1914 SAMD10 GMEB2 C20orf181 MIR941-3 ZBTB46 PCMTD2 RGS19 PRPF6 DNAJC5 RTEL1-TNFRSF6B ABHD16B OPRL1 ZNF512B MIR941-1 MIR6813 UCKL1-AS1 TCEA2 STMN3 RTEL1 UCKL1 ARFRP1 LINC00266-1
esv7227 Chr.20:63715550 - 63807161 on Build GRCh38 Loss SLC2A4RG ZGPAT LIME1 ZBTB46
nsv1059142 Chr.20:63731978 - 63798885 on Build GRCh38 Gain SLC2A4RG ZGPAT LIME1 ZBTB46
nsv586615 Chr.20:63734204 - 63741603 on Build GRCh38 Gain SLC2A4RG ZGPAT LIME1
nsv470563 Chr.20:63486715 - 63793308 on Build GRCh38 Loss EEF1A2 GMEB2 SLC2A4RG ZGPAT SRMS LIME1 ZBTB46 LOC100505771 PPDPF RTEL1-TNFRSF6B TNFRSF6B FNDC11 PTK6 STMN3 RTEL1 ARFRP1 HELZ2
esv2758806 Chr.20:63470996 - 63745014 on Build GRCh38 Gain+Loss EEF1A2 GMEB2 SLC2A4RG ZGPAT SRMS LIME1 ZBTB46 LOC100505771 PPDPF RTEL1-TNFRSF6B TNFRSF6B FNDC11 KCNQ2 PTK6 STMN3 RTEL1 ARFRP1 HELZ2

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s)

adaptive immune response
T cell receptor signaling pathway
B cell receptor signaling pathway

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline