This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02118653_cn
See other MEGF6 CNV Assays ›
Gene Symbol
MEGF6
Assay Reference Genome
Location

Chr.1:3488490 on build GRCh38
Cytoband
1p36.32
Assay ID Hs02118653_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

1953

Gene Name:

multiple EGF like domains 6

Gene Aliases:

EGFL3

Location:

Chr.1:3487942-3624757 on Build GRCh38

Assay Gene Location:

Within Exon 41
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MEGF6 NM_001409.3 37 6891 NP_001400.3
XM_006710406.3 34 6526 XP_006710469.1
XM_011540885.1 38 7196 XP_011539187.1
XM_011540886.1 37 7067 XP_011539188.1
XM_011540887.2 33 6404 XP_011539189.1
XM_017000533.1 37 7064 XP_016856022.1
AF086414.1 1 177
BM984873.1 1 560

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2758915 Chr.1:3372374 - 3632289 on Build GRCh38 Gain MEGF6 PRDM16 TPRG1L MIR551A WRAP73 ARHGEF16
nsv470687 Chr.1:3356499 - 3763210 on Build GRCh38 Loss MEGF6 PRDM16 TP73 TPRG1L MIR551A CCDC27 WRAP73 TP73-AS1 ARHGEF16
nsv1012423 Chr.1:3477686 - 4747449 on Build GRCh38 Gain MEGF6 LINC01346 TP73 TPRG1L WRAP73 AJAP1 LOC284661 TP73-AS1 LINC01134 CEP104 C1orf174 SMIM1 MIR551A CCDC27 DFFB LRRC47 ARHGEF16
dgv7n67 Chr.1:3484509 - 3489098 on Build GRCh38 Gain MEGF6
nsv951959 Chr.1:3051037 - 3812036 on Build GRCh38 Deletion MEGF6 TP73 LINC00982 TPRG1L WRAP73 TP73-AS1 PRDM16 SMIM1 MIR551A CCDC27 MIR4251 LRRC47 LOC105378604 ARHGEF16
esv3893545 Chr.1:3460878 - 3512044 on Build GRCh38 Loss MEGF6 ARHGEF16
nsv830203 Chr.1:3446374 - 3607752 on Build GRCh38 Loss MEGF6 MIR551A ARHGEF16
nsv545124 Chr.1:3451579 - 3488732 on Build GRCh38 Loss MEGF6 ARHGEF16
nsv299 Chr.1:3476928 - 3512705 on Build GRCh38 Insertion MEGF6 ARHGEF16

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

extracellular matrix protein

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline