This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02124048_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1785650 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02124048_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 10 2683 NP_001269467.1
NM_001282539.1 11 2780 NP_001269468.1
NM_002074.4 12 2829 NP_002065.1
XM_017001059.1 11 2733 XP_016856548.1
AI092255.1 1 372
AK122949.1 10 2467
BC004186.2 12 2712 AAH04186.1
BC005888.2 10 2480 AAH05888.1
BU684444.1 1 382
X04526.1 12 2716 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MMP23B SLC35E2B FNDC10 MIB2 CDK11B GNB1 MMP23A CDK11A SLC35E2 NADK
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss GNB1 MMP23A CDK11A SLC35E2 NADK
nsv951380 Chr.1:1785162 - 1791261 on Build GRCh38 Deletion GNB1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MMP23B SLC35E2B FNDC10 MIB2 CDK11B GNB1 MMP23A CDK11A SSU72 SLC35E2 NADK
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss PERM1 B3GALT6 LOC107985729 MIB2 CCNL2 ANKRD65 AGRN SLC35E2B OR4F29 MIR6859-2 GNB1 DDX11L1 RNF223 AURKAIP1 TMEM52 PRKCZ FAAP20 FNDC10 LOC100129534 LOC100288175 TMEM240 MTND1P23 LOC100130417 CALML6 LOC107985728 PUSL1 LINC00115 MMP23A KLHL17 MTND2P28 HES4 FAM41C SAMD11 MRPL20 LOC100133331 CDK11B LOC729737 CFAP74 MIR6727 LOC100288069 SSU72 MIR200B MIR200A MORN1 FAM87B ATAD3B TNFRSF4 WASH7P GABRD TAS1R3 LOC105378589 SKI CPSF3L SLC35E2 MIR6726 LOC100132287 LOC105378592 MXRA8 TNFRSF18 LOC100287934 FAM132A TTLL10 LOC101928626 MIR6808 MIR6723 VWA1 LOC102725121 LINC01342 C1orf159 OR4F16 OR4F5 TMEM88B LINC01128 CPTP LOC105378948 PLEKHN1 MIR6859-1 LOC107984841 LOC100506504 LOC148413 NOC2L ACAP3 MMP23B LOC105378591 SDF4 SCNN1D LOC284600 LOC105378947 CDK11A ATAD3C TTLL10-AS1 LOC105378949 ISG15 LOC102724312 UBE2J2 NADK LOC100134822 MIR1302-2 MIR429 ATAD3A FAM138A DVL1
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 GNB1 NADK
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain GNB1 NADK
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss GABRD CALML6 LOC107985729 MIB2 CCNL2 LOC105378589 MMP23A ANKRD65 SLC35E2 LOC148413 MMP23B SLC35E2B LOC105378591 CDK11A ATAD3C LOC105378949 MRPL20 CDK11B GNB1 LOC102724312 CFAP74 SSU72 NADK TMEM52 PRKCZ VWA1 FNDC10 ATAD3B ATAD3A TMEM240 TMEM88B

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline