This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02125216_cn
See other DHDDS CNV Assays ›
Gene Symbol
DHDDS
Assay Reference Genome
Location

Chr.1:26457843 on build GRCh38
Cytoband
1p36.11
Assay ID Hs02125216_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79947

Gene Name:

dehydrodolichyl diphosphate synthase subunit

Gene Aliases:

CIT, CPT, DS, HDS, RP59

Location:

Chr.1:26432282-26471306 on Build GRCh38

Assay Gene Location:

Within Exon 7
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DHDDS NM_001243564.1 6 643 NP_001230493.1
NM_001243565.1 6 628 NP_001230494.1
NM_001319959.1 7 768 NP_001306888.1
NM_024887.3 7 745 NP_079163.2
NM_205861.2 7 745 NP_995583.1
XM_006710912.2 7 804 XP_006710975.1
XM_006710913.2 7 683 XP_006710976.1
XM_006710914.2 7 680 XP_006710977.1
XM_006710915.2 6 615 XP_006710978.1
XM_006710916.2 7 794 XP_006710979.1
XM_006710917.2 7 747 XP_006710980.1
XM_006710918.2 7 703 XP_006710981.1
XM_011542183.2 7 780 XP_011540485.1
XM_011542184.2 7 801 XP_011540486.1
XM_011542185.2 6 699 XP_011540487.1
XM_011542186.2 6 699 XP_011540488.1
XM_017002379.1 7 683 XP_016857868.1
XM_017002380.1 7 780 XP_016857869.1
XM_017002381.1 7 794 XP_016857870.1
XM_017002382.1 7 703 XP_016857871.1
XM_017002383.1 6 692 XP_016857872.1
XM_017002384.1 6 645 XP_016857873.1
AB090852.1 6 606 BAC57588.1
AK023164.1 7 675 BAB14439.1
AK297134.1 5 563
AK308187.1 7 687
AK316485.1 6 586
BC003643.2 6 650 AAH03643.1
BC004117.2 6 570 AAH04117.1
BC034152.1 AAH34152.1
BX648507.1 7 770
GU727641.1 6 650

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
dolichyl diphosphate biosynthetic process
polyprenyltransferase activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0