This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02178074_cn
See other HES5 CNV Assays ›
Gene Symbol
HES5
Assay Reference Genome
Location

Chr.1:2529941 on build GRCh38
Cytoband
1p36.32
Assay ID Hs02178074_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

388585

Gene Name:

hes family bHLH transcription factor 5

Gene Aliases:

bHLHb38

Location:

Chr.1:2528745-2530554 on Build GRCh38

Assay Gene Location:

Within Exon 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
HES5 NM_001010926.3 2 206 NP_001010926.1
XM_005244751.4 2 515 XP_005244808.1
BC087840.1 1 305 AAH87840.1
DQ272660.1 2 206 ABB83829.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951557 Chr.1:2266062 - 2564661 on Build GRCh38 Deletion HES5 SKI PLCH2 MORN1 PEX10 LOC115110 LOC100129534 RER1 PANK4 TNFRSF14
nsv823920 Chr.1:2482305 - 2552402 on Build GRCh38 Gain HES5 PLCH2 LOC115110 PANK4
nsv818222 Chr.1:2492683 - 2553994 on Build GRCh38 Loss HES5 PLCH2 LOC115110 PANK4
nsv545080 Chr.1:2464013 - 2553994 on Build GRCh38 Loss HES5 PLCH2 LOC115110 PANK4
nsv545077 Chr.1:2428542 - 2622185 on Build GRCh38 Gain HES5 PLCH2 LOC100996583 LOC115110 MMEL1 PANK4 TNFRSF14 FAM213B
nsv470684 Chr.1:2394570 - 2930923 on Build GRCh38 Loss HES5 PLCH2 LOC100996583 PEX10 LOC115110 TTC34 MMEL1 RER1 PANK4 TNFRSF14 FAM213B
nsv824031 Chr.1:2520800 - 2551658 on Build GRCh38 Gain HES5 LOC115110 PANK4

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
transcription, DNA-templated
protein complex assembly
cell adhesion
Notch signaling pathway
smoothened signaling pathway
brain development
positive regulation of cell proliferation
oligodendrocyte development
telencephalon development
glial cell fate commitment
forebrain radial glial cell differentiation
neural tube development
central nervous system myelination
positive regulation of BMP signaling pathway
regulation of myelination
auditory receptor cell differentiation
positive regulation of tyrosine phosphorylation of Stat3 protein
auditory receptor cell fate determination
camera-type eye development
regulation of cell differentiation
regulation of auditory receptor cell differentiation
negative regulation of neuron differentiation
positive regulation of Notch signaling pathway
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
cell maturation
positive regulation of smooth muscle cell proliferation
astrocyte differentiation
negative regulation of astrocyte differentiation
negative regulation of oligodendrocyte differentiation
regulation of epithelial cell proliferation
regulation of neurogenesis
cartilage development
inner ear receptor stereocilium organization
comma-shaped body morphogenesis
S-shaped body morphogenesis
specification of loop of Henle identity
metanephric nephron tubule morphogenesis
establishment of epithelial cell polarity
neuronal stem cell population maintenance
negative regulation of stem cell differentiation
negative regulation of pro-B cell differentiation
negative regulation of forebrain neuron differentiation
negative regulation of inner ear receptor cell differentiation
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
chromatin binding
double-stranded DNA binding
transcription factor binding
protein dimerization activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline