This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02227718_cn
See other EIF4G3 CNV Assays ›
Gene Symbol
EIF4G3
Assay Reference Genome
Location

Chr.1:20807442 on build GRCh38
Cytoband
1p36.12
Assay ID Hs02227718_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8672

Gene Name:

eukaryotic translation initiation factor 4 gamma 3

Gene Aliases:

eIF-4G 3, eIF4G 3, eIF4GII

Location:

Chr.1:20806292-21177320 on Build GRCh38

Assay Gene Location:

Within Exon 43
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4G3 NM_001198801.1 35 5365 NP_001185730.1
NM_001198802.1 35 5275 NP_001185731.1
NM_003760.4 32 4891 NP_003751.2
XM_011542356.2 38 5965 XP_011540658.1
XM_011542361.2 37 5854 XP_011540663.1
XM_011542370.1 38 5968 XP_011540672.1
XM_011542371.1 36 5710 XP_011540673.1
XM_011542374.1 35 5677 XP_011540676.1
XM_011542375.2 35 7265 XP_011540677.1
XM_011542381.2 25 3351 XP_011540683.1
XM_017002672.1 39 7503 XP_016858161.1
XM_017002673.1 37 5947 XP_016858162.1
XM_017002674.1 37 5944 XP_016858163.1
XM_017002675.1 37 5935 XP_016858164.1
XM_017002676.1 36 5914 XP_016858165.1
XM_017002677.1 36 5791 XP_016858166.1
XM_017002678.1 37 5857 XP_016858167.1
XM_017002679.1 37 5854 XP_016858168.1
XM_017002680.1 34 4884 XP_016858169.1
XM_017002681.1 37 5851 XP_016858170.1
XM_017002682.1 36 5833 XP_016858171.1
XM_017002683.1 36 5833 XP_016858172.1
XM_017002684.1 37 7359 XP_016858173.1
XM_017002685.1 36 5824 XP_016858174.1
XM_017002686.1 36 5821 XP_016858175.1
XM_017002687.1 36 5821 XP_016858176.1
XM_017002688.1 33 4850 XP_016858177.1
XM_017002689.1 37 5821 XP_016858178.1
XM_017002690.1 36 5818 XP_016858179.1
XM_017002691.1 35 5803 XP_016858180.1
XM_017002692.1 35 5800 XP_016858181.1
XM_017002693.1 34 5671 XP_016858182.1
XM_017002694.1 36 5743 XP_016858183.1
XM_017002695.1 36 5740 XP_016858184.1
XM_017002696.1 35 5722 XP_016858185.1
XM_017002697.1 35 5719 XP_016858186.1
XM_017002698.1 35 5710 XP_016858187.1
XM_017002699.1 36 5710 XP_016858188.1
XM_017002700.1 34 5689 XP_016858189.1
XM_017002701.1 34 5686 XP_016858190.1
XM_017002702.1 34 5620 XP_016858191.1
XM_017002703.1 32 4781 XP_016858192.1
XM_017002704.1 36 5833 XP_016858193.1
XM_017002705.1 37 5128 XP_016858194.1
XM_017002706.1 36 5017 XP_016858195.1
XM_017002707.1 34 4960 XP_016858196.1
XM_017002708.1 33 4849 XP_016858197.1
XM_017002709.1 34 5565 XP_016858198.1
AB209119.1 35 5234 BAD92356.1
AF012072.2 32 4891 AAC02903.2
AK294883.1 26 4138
AK302873.1 23 3132
BC094683.1 30 3898 AAH94683.1
BC136643.1 32 4814
Z34918.1 16 1977 CAA84397.1

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-4gzsq:80/100.66.74.9:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0