This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02308711_cn
See other UBE2J2 CNV Assays ›
Gene Symbol
UBE2J2
Assay Reference Genome
Location

Chr.1:1263397 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02308711_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

118424

Gene Name:

ubiquitin conjugating enzyme E2 J2

Gene Aliases:

NCUBE-2, NCUBE2, PRO2121

Location:

Chr.1:1253912-1273854 on Build GRCh38

Assay Gene Location:

Overlaps Intron 4 - Exon 5
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
UBE2J2 NM_058167.2 NP_477515.2
NM_194315.1 NP_919296.1
NM_194457.1 NP_919439.1
NM_194458.1 NP_919440.1
XM_005244718.3 XP_005244775.1
XM_005244719.4 XP_005244776.1
XM_006710333.3 XP_006710396.1
XM_011540613.2 XP_011538915.1
XM_011540614.2 XP_011538916.1
XM_011540616.2 XP_011538918.1
XM_011540617.2 XP_011538919.1
XM_017000238.1 XP_016855727.1
XM_017000239.1 XP_016855728.1
AF296658.1 AAK52609.1
AK056065.1 BAB71086.1
AK075017.1 BAC11355.1
AK291935.1
AL555293.3
BC094777.1 AAH94777.1
BC104890.1 AAI04891.1
BC113430.1
BC143657.1
BE382927.1
BG499172.1
BG717336.1
BM544827.1
BM765278.1
BM794599.1
BM926567.1
BQ881730.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv817616 Chr.1:1257135 - 1306704 on Build GRCh38 Loss UBE2J2 MIR6726 ACAP3 SCNN1D
nsv1160772 Chr.1:1027585 - 1333467 on Build GRCh38 Duplication TTLL10-AS1 RNF223 TNFRSF18 C1orf159 ACAP3 FAM132A TNFRSF4 UBE2J2 MIR200A TAS1R3 AGRN SCNN1D LINC01342 MIR6727 B3GALT6 MIR200B PUSL1 MIR429 MIR6726 LOC105378948 CPSF3L LOC100288175 SDF4 TTLL10 CPTP
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss TTLL10-AS1 TMEM88B MMP23B SSU72 AURKAIP1 RNF223 TNFRSF18 FNDC10 SLC35E2 FAM132A TNFRSF4 NADK NOC2L UBE2J2 TMEM240 LOC100130417 TAS1R3 FAM41C VWA1 MXRA8 SAMD11 MRPL20 AGRN LINC01342 SLC35E2B MIR6727 CDK11A FAM87B LOC148413 B3GALT6 PUSL1 ATAD3C LOC107985728 ATAD3A LINC00115 LOC100288175 TTLL10 CPTP ISG15 C1orf159 LOC107985729 ACAP3 MIB2 CDK11B CCNL2 MIR6808 ANKRD65 MMP23A PLEKHN1 LINC01128 MIR200A LOC102724312 LOC100287934 HES4 LOC284600 KLHL17 SCNN1D ATAD3B MIR200B MIR429 MIR6726 LOC105378948 CPSF3L PERM1 SDF4 DVL1 LOC100288069
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss TMEM88B SSU72 AURKAIP1 RNF223 PRKCZ SLC35E2 LOC100132287 LOC729737 MIR1302-2 LOC100130417 TAS1R3 LOC105378947 VWA1 MXRA8 SAMD11 TMEM52 SLC35E2B LOC100506504 CDK11A LOC148413 PUSL1 LOC107984841 ATAD3A LINC00115 TTLL10 ISG15 C1orf159 LOC107985729 ACAP3 MIB2 MIR6859-2 CCNL2 MIR6808 ANKRD65 MMP23A FAAP20 PLEKHN1 LINC01128 MIR200A LOC102724312 LOC100287934 LOC284600 GNB1 SCNN1D LOC105378592 OR4F29 MORN1 CFAP74 LOC105378589 MIR6726 LOC105378948 DVL1 LOC100288069 MTND2P28 OR4F5 TTLL10-AS1 MMP23B TNFRSF18 LOC102725121 FNDC10 FAM132A FAM138A TNFRSF4 NADK NOC2L UBE2J2 WASH7P TMEM240 LOC100129534 FAM41C MRPL20 AGRN MTND1P23 LINC01342 LOC100133331 MIR6727 FAM87B B3GALT6 ATAD3C LOC107985728 MIR6723 LOC100288175 CPTP LOC101928626 OR4F16 LOC105378949 CDK11B DDX11L1 CALML6 MIR6859-1 HES4 KLHL17 GABRD LOC100134822 SKI ATAD3B MIR200B MIR429 LOC105378591 CPSF3L PERM1 SDF4
nsv544965 Chr.1:1217251 - 1306704 on Build GRCh38 Loss B3GALT6 UBE2J2 MIR6726 ACAP3 SCNN1D SDF4 FAM132A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss TTLL10-AS1 RNF223 TNFRSF18 C1orf159 FAM132A TNFRSF4 NOC2L UBE2J2 PLEKHN1 LINC01128 LOC100130417 MIR200A FAM41C HES4 LOC284600 KLHL17 SAMD11 AGRN LINC01342 FAM87B B3GALT6 MIR200B MIR429 LOC107985728 LOC105378948 LINC00115 PERM1 LOC100288175 SDF4 TTLL10 ISG15
nsv950452 Chr.1:1106921 - 1637538 on Build GRCh38 Deletion TTLL10-AS1 TMEM88B MMP23B SSU72 AURKAIP1 TNFRSF18 C1orf159 LOC107985729 ACAP3 FNDC10 MIB2 FAM132A TNFRSF4 CDK11B CCNL2 MIR6808 ANKRD65 UBE2J2 TMEM240 MIR200A TAS1R3 LOC102724312 VWA1 MXRA8 MRPL20 SCNN1D LINC01342 ATAD3B MIR6727 LOC148413 B3GALT6 MIR200B PUSL1 ATAD3C MIR429 MIR6726 CPSF3L ATAD3A SDF4 TTLL10 CPTP DVL1
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain TTLL10-AS1 TMEM88B AURKAIP1 RNF223 TNFRSF18 C1orf159 ACAP3 FAM132A TNFRSF4 CCNL2 MIR6808 ANKRD65 NOC2L UBE2J2 PLEKHN1 LOC100130417 MIR200A TAS1R3 HES4 LOC284600 KLHL17 MXRA8 SAMD11 MRPL20 AGRN SCNN1D LINC01342 MIR6727 LOC148413 B3GALT6 MIR200B PUSL1 MIR429 MIR6726 LOC107985728 LOC105378948 CPSF3L PERM1 LOC100288175 SDF4 TTLL10 CPTP DVL1 ISG15
nsv1003578 Chr.1:1068249 - 1282706 on Build GRCh38 Gain MIR200A TTLL10-AS1 RNF223 TNFRSF18 C1orf159 SCNN1D FAM132A LINC01342 TNFRSF4 B3GALT6 MIR200B UBE2J2 MIR429 LOC105378948 SDF4 TTLL10
nsv544947 Chr.1:1070426 - 1306704 on Build GRCh38 Loss MIR200A TTLL10-AS1 RNF223 TNFRSF18 C1orf159 ACAP3 SCNN1D FAM132A LINC01342 TNFRSF4 B3GALT6 MIR200B UBE2J2 MIR429 MIR6726 SDF4 TTLL10
nsv469848 Chr.1:1146082 - 1322235 on Build GRCh38 Gain MIR200A TTLL10-AS1 TNFRSF18 ACAP3 SCNN1D FAM132A TNFRSF4 MIR6727 B3GALT6 MIR200B UBE2J2 PUSL1 MIR429 MIR6726 CPSF3L SDF4 TTLL10

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-65nqn:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline