Assay Details
Target Gene Details
Entrez Gene ID: | 65999 |
Gene Name: | leucine rich repeat containing 61 |
Gene Aliases: |
HSPC295 |
Location: |
Chr.7:150322639-150338156 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRRC61 | NM_001142928.1 | NP_001136400.1 | ||
| NM_023942.2 | NP_076431.1 | |||
| XM_006716095.2 | XP_006716158.1 | |||
| AK025869.1 | ||||
| AK055954.1 | ||||
| AK098061.1 | ||||
| BC001354.1 | AAH01354.1 |
Target Gene Details
Entrez Gene ID: | 113763 |
Gene Name: | ZBED6 C-terminal like |
Gene Aliases: |
C7orf29 |
Location: |
Chr.7:150329849-150332721 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZBED6CL | NM_138434.2 | 1 | 264 | NP_612443.1 |
| AL832660.1 | 1 | 326 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1034409 | Chr.7:150147550 - 150749983 on Build GRCh38 | Gain |
|
| nsv1015582 | Chr.7:150270897 - 150588016 on Build GRCh38 | Gain |
|
| nsv508490 | Chr.7:150308979 - 150354152 on Build GRCh38 | Deletion |
|
| nsv951645 | Chr.7:150326212 - 150342411 on Build GRCh38 | Deletion |
|
| nsv608990 | Chr.7:150264185 - 151010400 on Build GRCh38 | Gain |
|
| nsv521407 | Chr.7:150323206 - 150421502 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs114925535] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map