Assay Details
Target Gene Details
Entrez Gene ID: | 161394 |
Gene Name: | sterile alpha motif domain containing 15 |
Gene Aliases: |
C14orf174, FAM15A |
Location: |
Chr.14:77377419-77391244 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD15 | NM_001010860.1 | NP_001010860.1 | ||
| AK093282.1 | ||||
| AK292571.1 | ||||
| BC104834.1 | 1 | 1697 | AAI04835.1 | |
| BC104836.1 | 1 | 1697 | AAI04837.1 |

Genomic Map