This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02507223_cn
See other C1ORF167 CNV Assays ›
Gene Symbol
C1orf167
Assay Reference Genome
Location

Chr.1:11787834 on build GRCh38
Cytoband
1p36.22
Assay ID Hs02507223_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

284498

Gene Name:

chromosome 1 open reading frame 167

Gene Aliases:

-

Location:

Chr.1:11762188-11789585 on Build GRCh38

Assay Gene Location:

Overlaps Intron 23 - Exon 24
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
C1orf167 NM_001010881.1 NP_001010881.1
XM_011541269.2 XP_011539571.2
XM_011541271.2 XP_011539573.2
XM_011541272.2 XP_011539574.1
XM_011541274.2 XP_011539576.2
XM_011541275.2 XP_011539577.2
XM_011541276.2 XP_011539578.1
XM_011541277.2 XP_011539579.1
XM_011541278.2 XP_011539580.1
XM_011541280.2 XP_011539582.2
XM_011541281.2 XP_011539583.2
XM_017001062.1 XP_016856551.1
XM_017001063.1 XP_016856552.1
XM_017001064.1 XP_016856553.1
AF398935.1
AL833926.1 CAD38782.1
AL834308.1 CAD38978.2
BC031692.1

Target Gene Details

Entrez Gene ID:

4524

Gene Name:

methylenetetrahydrofolate reductase (NAD(P)H)

Gene Aliases:

-

Location:

Chr.1:11785730-11806103 on Build GRCh38

Assay Gene Location:

Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MTHFR NM_005957.4 12 5046 NP_005948.3
AB209113.1 12 3846 BAD92350.1
BC053509.1 12 4868 AAH53509.1

Back To Top

More Information


Set Membership:

Intragenic Exonic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
cellular amino acid metabolic process
methionine metabolic process
blood circulation
regulation of histone methylation
response to vitamin B2
tetrahydrofolate interconversion
response to drug
response to amino acid
S-adenosylmethionine metabolic process
folic acid metabolic process
homocysteine metabolic process
response to folic acid
oxidation-reduction process
response to interleukin-1
heterochromatin maintenance
methylenetetrahydrofolate reductase (NAD(P)H) activity
protein complex binding
flavin adenine dinucleotide binding
NADP binding
modified amino acid binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline