This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02507647_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1786298 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02507647_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 10 2035 NP_001269467.1
NM_001282539.1 11 2132 NP_001269468.1
NM_002074.4 12 2181 NP_002065.1
XM_017001059.1 11 2085 XP_016856548.1
XM_017001060.1 12 1951 XP_016856549.1
XM_017001061.1 11 2124 XP_016856550.1
AF070597.1 4 1150 AAC28652.1
AF070603.1 6 1389 AAC28655.1
AK122949.1 10 1819
BC004186.2 12 2064 AAH04186.1
BC005888.2 10 1832 AAH05888.1
X04526.1 12 2068 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MMP23A NADK CDK11B CDK11A SLC35E2B SLC35E2 MMP23B FNDC10 GNB1 MIB2
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss MMP23A NADK CDK11A SLC35E2 GNB1
nsv951380 Chr.1:1785162 - 1791261 on Build GRCh38 Deletion GNB1
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss CDK11B MIR6859-1 TMEM88B B3GALT6 MTND1P23 SKI LINC01342 LOC105378947 LOC107984841 SLC35E2B MTND2P28 OR4F16 FAM87B MIR200A ACAP3 GABRD OR4F5 UBE2J2 NADK ISG15 PLEKHN1 MXRA8 LOC105378948 MORN1 CPSF3L FAM132A FNDC10 LOC105378591 LOC100288069 LOC284600 MIR1302-2 LOC100133331 LOC729737 PERM1 LOC107985728 CDK11A TNFRSF18 MIR6808 RNF223 MMP23B ANKRD65 SSU72 AURKAIP1 LOC105378589 CPTP TTLL10 TAS1R3 MIR200B LOC107985729 LINC01128 SAMD11 GNB1 MIR6723 DDX11L1 CALML6 KLHL17 ATAD3B SCNN1D ATAD3C VWA1 SDF4 AGRN CFAP74 OR4F29 TMEM52 LOC100130417 LOC100506504 CCNL2 LOC100129534 C1orf159 MIR6726 DVL1 WASH7P LOC102725121 MIR6727 ATAD3A LOC100287934 PUSL1 LINC00115 MMP23A LOC102724312 TNFRSF4 FAM138A TTLL10-AS1 MIR429 FAM41C HES4 LOC105378949 NOC2L MIB2 LOC148413 TMEM240 MRPL20 FAAP20 SLC35E2 LOC100134822 LOC101928626 LOC100132287 PRKCZ LOC100288175 MIR6859-2 LOC105378592
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MMP23A NADK CDK11B CDK11A SLC35E2B SLC35E2 MMP23B SSU72 FNDC10 GNB1 MIB2
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication NADK LOC105378949 GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss CDK11B ATAD3B TMEM88B ATAD3C VWA1 FNDC10 LOC105378591 CFAP74 CDK11A SLC35E2B TMEM52 MMP23B ANKRD65 SSU72 CCNL2 LOC105378949 LOC105378589 MIB2 LOC148413 TMEM240 MRPL20 LOC107985729 GABRD SLC35E2 ATAD3A PRKCZ GNB1 MMP23A NADK LOC102724312 CALML6

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-rdsp8:80/100.66.79.164:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline