This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02536428_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:945483 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02536428_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Overlaps Intron 17 - Exon 17
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK022756.1 BAB14230.1
AK024284.1
AK092491.1 BAC03905.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain C1orf159 AGRN LOC284600 LOC100130417 LOC105378948 LOC107985728 SAMD11 ISG15 PERM1 PLEKHN1 LOC100288175 NOC2L RNF223 KLHL17 HES4
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss GNB1 CDK11A OR4F5 LOC729737 ANKRD65 TMEM88B LOC107984841 LOC105378589 LOC102725121 LOC105378949 LOC101928626 TTLL10-AS1 LOC100288175 MIR6727 SSU72 SLC35E2 B3GALT6 WASH7P MORN1 PERM1 DVL1 MIR1302-2 TMEM52 CDK11B LINC01128 NADK HES4 C1orf159 LOC100133331 ISG15 CPSF3L MIB2 MIR429 LOC100129534 AURKAIP1 FAM132A TMEM240 VWA1 MTND2P28 MMP23A CFAP74 LOC102724312 LOC100130417 SAMD11 ACAP3 LINC01342 TAS1R3 LINC00115 FAM41C PLEKHN1 MMP23B FAAP20 LOC107985729 MTND1P23 UBE2J2 MIR6723 LOC107985728 LOC100506504 LOC100287934 LOC148413 FAM138A SLC35E2B LOC105378948 FNDC10 MIR6726 SCNN1D PRKCZ NOC2L RNF223 KLHL17 AGRN OR4F16 LOC100132287 PUSL1 MIR200A MIR6859-1 TTLL10 LOC105378592 LOC100134822 LOC105378947 CCNL2 MIR6859-2 CPTP ATAD3B GABRD MRPL20 LOC284600 LOC100288069 MXRA8 LOC105378591 SDF4 MIR200B CALML6 TNFRSF18 FAM87B SKI ATAD3A ATAD3C DDX11L1 TNFRSF4 OR4F29 MIR6808
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion C1orf159 AGRN LOC284600 LOC100130417 LOC105378948 LOC107985728 SAMD11 ISG15 PERM1 PLEKHN1 LOC100288175 NOC2L RNF223 KLHL17 HES4
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion LOC284600 PLEKHN1 LOC100130417 LOC107985728 SAMD11 NOC2L KLHL17 PERM1
dgv9n54 Chr.1:925754 - 952840 on Build GRCh38 Loss SAMD11 NOC2L
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain C1orf159 AGRN LOC107985728 MIR200A TTLL10 ISG15 MIR429 TTLL10-AS1 LOC100288175 LOC284600 LOC100130417 LOC105378948 SDF4 SAMD11 MIR200B TNFRSF18 LINC01342 PERM1 FAM41C PLEKHN1 TNFRSF4 NOC2L RNF223 KLHL17 HES4
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain C1orf159 AGRN OR4F16 MIR6723 LOC100133331 LOC107985728 ISG15 LOC107984841 LOC101928626 LOC100287934 LOC100288175 LOC105378947 MTND2P28 LOC284600 LOC100288069 LOC100130417 LOC105378948 SAMD11 PERM1 LINC00115 FAM87B FAM41C PLEKHN1 NOC2L RNF223 KLHL17 LINC01128 HES4 MTND1P23
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss CDK11A LOC107985728 ANKRD65 TMEM88B LOC100287934 TTLL10-AS1 LOC100288175 MIR6727 SSU72 LOC148413 SLC35E2 B3GALT6 SLC35E2B LOC105378948 FNDC10 MIR6726 PERM1 SCNN1D DVL1 NOC2L RNF223 CDK11B KLHL17 LINC01128 NADK HES4 C1orf159 AGRN PUSL1 MIR200A TTLL10 ISG15 CPSF3L MIB2 MIR429 AURKAIP1 FAM132A CCNL2 TMEM240 CPTP VWA1 ATAD3B MRPL20 MMP23A LOC102724312 LOC284600 LOC100288069 MXRA8 LOC100130417 SDF4 SAMD11 MIR200B ACAP3 TNFRSF18 LINC01342 TAS1R3 LINC00115 FAM87B FAM41C PLEKHN1 ATAD3A ATAD3C MMP23B TNFRSF4 LOC107985729 MIR6808 UBE2J2
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss C1orf159 AGRN LOC107985728 MIR200A TTLL10 ISG15 MIR429 FAM132A TTLL10-AS1 LOC100288175 B3GALT6 LOC284600 LOC100130417 LOC105378948 SDF4 SAMD11 MIR200B TNFRSF18 LINC01342 PERM1 LINC00115 FAM87B FAM41C PLEKHN1 TNFRSF4 NOC2L RNF223 KLHL17 LINC01128 HES4 UBE2J2
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain C1orf159 AGRN LOC105378948 SAMD11 ISG15 LINC01342 PERM1 PLEKHN1 LOC100288175 NOC2L RNF223 KLHL17 HES4
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain C1orf159 AGRN PUSL1 LOC107985728 MIR200A TTLL10 ANKRD65 TMEM88B ISG15 CPSF3L MIR429 AURKAIP1 FAM132A TTLL10-AS1 LOC100288175 CCNL2 MIR6727 CPTP LOC148413 MRPL20 B3GALT6 LOC284600 MXRA8 LOC100130417 LOC105378948 SDF4 SAMD11 MIR200B ACAP3 TNFRSF18 LINC01342 TAS1R3 MIR6726 PERM1 SCNN1D DVL1 PLEKHN1 TNFRSF4 NOC2L RNF223 KLHL17 HES4 MIR6808 UBE2J2
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss AGRN LOC284600 PLEKHN1 LOC100130417 LOC107985728 SAMD11 NOC2L KLHL17 ISG15 HES4 PERM1
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion C1orf159 AGRN LOC105378948 SDF4 MIR200A SAMD11 MIR200B TTLL10 TNFRSF18 ISG15 LINC01342 PERM1 MIR429 PLEKHN1 TTLL10-AS1 LOC100288175 TNFRSF4 NOC2L RNF223 KLHL17 HES4
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain FAM41C LOC284600 PLEKHN1 LOC100130417 LOC107985728 SAMD11 NOC2L KLHL17 LINC01128 HES4 PERM1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-2x6nv:80/100.66.78.26:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline