This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02547397_cn
See other NOC2L CNV Assays ›
Gene Symbol
NOC2L
Assay Reference Genome
Location

Chr.1:948633 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02547397_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

26155

Gene Name:

NOC2 like nucleolar associated transcriptional repressor

Gene Aliases:

NET15, NET7, NIR, PPP1R112

Location:

Chr.1:944203-959299 on Build GRCh38

Assay Gene Location:

Overlaps Intron 12 - Exon 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
NOC2L NM_015658.3 NP_056473.2
AK022756.1 BAB14230.1
AK024284.1
AK225239.1
AK315080.1
AL050019.1 CAB43240.2
BC003555.1 AAH03555.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2762302 Chr.1:914637 - 1106320 on Build GRCh38 Gain ISG15 PLEKHN1 SAMD11 LOC100130417 LOC105378948 AGRN RNF223 NOC2L HES4 PERM1 C1orf159 LOC100288175 LOC284600 KLHL17 LOC107985728
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss VWA1 MIR6723 LOC148413 OR4F16 LOC100133331 NOC2L LOC102724312 MTND2P28 LOC100287934 MTND1P23 PRKCZ FAM87B TTLL10-AS1 LOC107985728 LOC101928626 LOC105378591 LOC100506504 TMEM52 MIR6808 NADK LOC107985729 PERM1 LOC105378947 FAM132A FAM41C ACAP3 MIR429 ATAD3B MIR6859-2 OR4F5 CDK11A TAS1R3 LOC100130417 LOC105378948 TNFRSF4 MXRA8 MIR6859-1 TMEM240 FNDC10 LOC105378589 MIR6726 LOC105378592 CALML6 LOC102725121 WASH7P LINC01128 CFAP74 SCNN1D TTLL10 B3GALT6 SLC35E2B PUSL1 UBE2J2 FAAP20 LOC100134822 CPSF3L C1orf159 MIR6727 LOC284600 CCNL2 MIR200B CDK11B SDF4 OR4F29 DDX11L1 FAM138A LINC01342 TMEM88B ATAD3A MMP23A AURKAIP1 MRPL20 ISG15 ATAD3C MMP23B SAMD11 SKI SSU72 AGRN LOC100129534 LOC107984841 RNF223 LOC100132287 TNFRSF18 LOC729737 MIR1302-2 MIB2 SLC35E2 ANKRD65 PLEKHN1 DVL1 GABRD LOC105378949 HES4 LINC00115 GNB1 LOC100288175 LOC100288069 CPTP KLHL17 MORN1 MIR200A
nsv950451 Chr.1:899421 - 1094520 on Build GRCh38 Deletion ISG15 PLEKHN1 SAMD11 LOC100130417 LOC105378948 AGRN RNF223 NOC2L HES4 PERM1 C1orf159 LOC100288175 LOC284600 KLHL17 LOC107985728
nsv509035 Chr.1:891406 - 985724 on Build GRCh38 Insertion PERM1 PLEKHN1 LOC284600 SAMD11 LOC100130417 KLHL17 NOC2L LOC107985728
dgv9n54 Chr.1:925754 - 952840 on Build GRCh38 Loss SAMD11 NOC2L
nsv428334 Chr.1:874371 - 1220569 on Build GRCh38 Gain ISG15 SAMD11 LOC100130417 LOC105378948 AGRN TNFRSF4 RNF223 NOC2L TNFRSF18 C1orf159 LOC284600 MIR200B TTLL10-AS1 SDF4 LOC107985728 PLEKHN1 HES4 PERM1 LINC01342 LOC100288175 KLHL17 TTLL10 FAM41C MIR200A MIR429
dgv5n100 Chr.1:585989 - 1114424 on Build GRCh38 Gain MIR6723 ISG15 SAMD11 LOC100130417 LOC105378948 OR4F16 AGRN LOC100133331 LOC107984841 RNF223 NOC2L MTND2P28 LOC100287934 MTND1P23 C1orf159 FAM87B LOC284600 LOC107985728 LOC101928626 PLEKHN1 HES4 LINC00115 PERM1 LINC01128 LOC100288175 LOC100288069 LOC105378947 KLHL17 FAM41C
nsv10161 Chr.1:776731 - 1777210 on Build GRCh38 Gain+Loss VWA1 SLC35E2B PUSL1 UBE2J2 LOC148413 NOC2L LOC102724312 LOC100287934 CPSF3L C1orf159 FAM87B MIR6727 LOC284600 CCNL2 MIR200B TTLL10-AS1 CDK11B SDF4 LOC107985728 MIR6808 NADK LOC107985729 PERM1 LINC01342 TMEM88B FAM132A ATAD3A FAM41C MMP23A ACAP3 MIR429 AURKAIP1 MRPL20 ATAD3B ISG15 CDK11A ATAD3C MMP23B TAS1R3 SAMD11 LOC100130417 LOC105378948 SSU72 AGRN TNFRSF4 RNF223 MXRA8 TNFRSF18 TMEM240 FNDC10 MIB2 SLC35E2 ANKRD65 PLEKHN1 MIR6726 DVL1 HES4 LINC00115 LINC01128 LOC100288175 LOC100288069 CPTP KLHL17 SCNN1D TTLL10 B3GALT6 MIR200A
nsv517709 Chr.1:817186 - 1275912 on Build GRCh38 Gain+Loss ISG15 UBE2J2 SAMD11 LOC100130417 LOC105378948 AGRN TNFRSF4 RNF223 NOC2L TNFRSF18 C1orf159 FAM87B LOC284600 MIR200B TTLL10-AS1 SDF4 LOC107985728 PLEKHN1 HES4 LINC00115 PERM1 LINC01128 LINC01342 LOC100288175 KLHL17 TTLL10 FAM132A FAM41C B3GALT6 MIR200A MIR429
nsv1013524 Chr.1:939510 - 1161955 on Build GRCh38 Gain ISG15 PLEKHN1 SAMD11 LOC105378948 AGRN RNF223 NOC2L HES4 PERM1 C1orf159 LINC01342 LOC100288175 KLHL17
dgv2n67 Chr.1:877618 - 1426500 on Build GRCh38 Gain AURKAIP1 MRPL20 PUSL1 ISG15 UBE2J2 LOC148413 TAS1R3 SAMD11 LOC100130417 LOC105378948 AGRN TNFRSF4 RNF223 NOC2L MXRA8 TNFRSF18 CPSF3L C1orf159 MIR6727 LOC284600 CCNL2 MIR200B TTLL10-AS1 ANKRD65 SDF4 LOC107985728 PLEKHN1 MIR6726 DVL1 MIR6808 HES4 PERM1 LINC01342 LOC100288175 CPTP KLHL17 TMEM88B SCNN1D TTLL10 FAM132A B3GALT6 MIR200A ACAP3 MIR429
nsv544895 Chr.1:917392 - 1054900 on Build GRCh38 Loss PERM1 ISG15 PLEKHN1 LOC284600 SAMD11 LOC100130417 KLHL17 AGRN NOC2L HES4 LOC107985728
nsv1160644 Chr.1:939652 - 1220663 on Build GRCh38 Deletion ISG15 PLEKHN1 SAMD11 LOC105378948 AGRN TNFRSF4 RNF223 NOC2L HES4 PERM1 TNFRSF18 C1orf159 LINC01342 LOC100288175 KLHL17 TTLL10 MIR200B TTLL10-AS1 SDF4 MIR200A MIR429
nsv832980 Chr.1:848279 - 1007037 on Build GRCh38 Gain PERM1 PLEKHN1 LINC01128 LOC284600 SAMD11 LOC100130417 KLHL17 NOC2L HES4 FAM41C LOC107985728

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
negative regulation of B cell apoptotic process
transcription, DNA-templated
apoptotic process
chromatin assembly
nucleolus to nucleoplasm transport
cellular response to UV
negative regulation of histone acetylation
ribosomal large subunit biogenesis
regulation of signal transduction by p53 class mediator
negative regulation of intrinsic apoptotic signaling pathway
chromatin binding
transcription corepressor activity
protein binding
nucleosome binding
histone binding
poly(A) RNA binding
repressing transcription factor binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-794c4b9cd6-8pzg4:80/100.66.75.138:80.
git-commit: 7e8dd63fde7d4486b98ef9ac5cf4e365aa842ffe
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.52.0-Offline