This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02558493_cn
See other FAM3B CNV Assays ›
Gene Symbol
FAM3B
Assay Reference Genome
Location

Chr.21:41316869 on build GRCh38
Cytoband
21q22.3
Assay ID Hs02558493_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

54097

Gene Name:

family with sequence similarity 3 member B

Gene Aliases:

2-21, C21orf11, C21orf76, ORF9, PANDER, PRED44

Location:

Chr.21:41304229-41357727 on Build GRCh38

Assay Gene Location:

Overlaps Exon 2 - Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FAM3B NM_058186.3 NP_478066.3
NM_206964.1 NP_996847.1
XM_011529649.2 XP_011527951.1
AF375989.1 AAL34495.1
AF494379.1 AAM94280.1
AJ409094.1 CAC83974.1
AK125528.1
AK302095.1
AY040086.1 AAK74134.1
AY358459.1 AAQ88824.1
BC057829.1 AAH57829.1
BI712971.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
esv2723486 Chr.21:41316771 - 41318499 on Build GRCh38 Deletion FAM3B
nsv834102 Chr.21:41286267 - 41494006 on Build GRCh38 Loss MX1 TMPRSS2 FAM3B MX2
nsv1057981 Chr.21:41290295 - 41422831 on Build GRCh38 Loss MX1 FAM3B MX2
esv3647058 Chr.21:41316747 - 41318377 on Build GRCh38 Loss FAM3B

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

antimicrobial response protein

Gene Ontology Categories:

Function(s) Process(es)

apoptotic process
insulin secretion
cytokine activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.47.0-Offline