This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02596592_cn
See other EIF4G3 CNV Assays ›
Gene Symbol
EIF4G3
Assay Reference Genome
Location

Chr.1:20807187 on build GRCh38
Cytoband
1p36.12
Assay ID Hs02596592_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

8672

Gene Name:

eukaryotic translation initiation factor 4 gamma 3

Gene Aliases:

eIF-4G 3, eIF4G 3, eIF4GII

Location:

Chr.1:20806292-21177320 on Build GRCh38

Assay Gene Location:

Within Exon 43
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
EIF4G3 NM_001198801.1 35 5620 NP_001185730.1
NM_001198802.1 35 5530 NP_001185731.1
NM_003760.4 32 5146 NP_003751.2
XM_011542356.2 38 6220 XP_011540658.1
XM_011542361.2 37 6109 XP_011540663.1
XM_011542370.1 38 6223 XP_011540672.1
XM_011542371.1 36 5965 XP_011540673.1
XM_011542374.1 35 5932 XP_011540676.1
XM_011542375.2 35 7520 XP_011540677.1
XM_011542381.2 25 3606 XP_011540683.1
XM_017002672.1 39 7758 XP_016858161.1
XM_017002673.1 37 6202 XP_016858162.1
XM_017002674.1 37 6199 XP_016858163.1
XM_017002675.1 37 6190 XP_016858164.1
XM_017002676.1 36 6169 XP_016858165.1
XM_017002677.1 36 6046 XP_016858166.1
XM_017002678.1 37 6112 XP_016858167.1
XM_017002679.1 37 6109 XP_016858168.1
XM_017002680.1 34 5139 XP_016858169.1
XM_017002681.1 37 6106 XP_016858170.1
XM_017002682.1 36 6088 XP_016858171.1
XM_017002683.1 36 6088 XP_016858172.1
XM_017002684.1 37 7614 XP_016858173.1
XM_017002685.1 36 6079 XP_016858174.1
XM_017002686.1 36 6076 XP_016858175.1
XM_017002687.1 36 6076 XP_016858176.1
XM_017002688.1 33 5105 XP_016858177.1
XM_017002689.1 37 6076 XP_016858178.1
XM_017002690.1 36 6073 XP_016858179.1
XM_017002691.1 35 6058 XP_016858180.1
XM_017002692.1 35 6055 XP_016858181.1
XM_017002693.1 34 5926 XP_016858182.1
XM_017002694.1 36 5998 XP_016858183.1
XM_017002695.1 36 5995 XP_016858184.1
XM_017002696.1 35 5977 XP_016858185.1
XM_017002697.1 35 5974 XP_016858186.1
XM_017002698.1 35 5965 XP_016858187.1
XM_017002699.1 36 5965 XP_016858188.1
XM_017002700.1 34 5944 XP_016858189.1
XM_017002701.1 34 5941 XP_016858190.1
XM_017002702.1 34 5875 XP_016858191.1
XM_017002703.1 32 5036 XP_016858192.1
XM_017002704.1 36 6088 XP_016858193.1
XM_017002705.1 37 5383 XP_016858194.1
XM_017002706.1 36 5272 XP_016858195.1
XM_017002707.1 34 5215 XP_016858196.1
XM_017002708.1 33 5104 XP_016858197.1
XM_017002709.1 34 5820 XP_016858198.1
AB209119.1 35 5489 BAD92356.1
AF012072.2 32 5146 AAC02903.2
AK302873.1 23 3387
BC094683.1 30 4153 AAH94683.1
BC136643.1

Back To Top

More Information


Set Membership:

Intragenic Exonic non-DGV Variation

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-hrdnk:80/100.66.78.145:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0