This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02638150_cn
See other ESRRA CNV Assays ›
Gene Symbol
ESRRA
Assay Reference Genome
Location

Chr.11:64316730 on build GRCh38
Cytoband
11q13.1
Assay ID Hs02638150_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2101

Gene Name:

estrogen related receptor alpha

Gene Aliases:

ERR1, ERRa, ERRalpha, ESRL1, NR3B1

Location:

Chr.11:64305528-64316743 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 8
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
ESRRA NM_001282450.1 NP_001269379.1
NM_001282451.1 NP_001269380.1
NM_004451.4 NP_004442.3
XM_017017313.1 XP_016872802.1
BC007915.2 AAH07915.2
BC011528.1 AAH11528.1
BC033701.1 AAH33701.1
BC063795.1
BC073956.1 AAH73956.1
BC092470.1 AAH92470.1
L38487.1 AAB17015.1

Target Gene Details

Entrez Gene ID:

51504

Gene Name:

tRNA methyltransferase 11-2 homolog (S. cerevisiae)

Gene Aliases:

HSPC152, HSPC170, TRM112, TRMT11-2, hTrm112

Location:

Chr.11:64316492-64318084 on Build GRCh38

Assay Gene Location:

Within Exon 3
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
TRMT112 NM_001286082.1 4 1052 NP_001273011.1
NM_001286084.1 3 1156 NP_001273013.1
NM_016404.2 4 1067 NP_057488.1
AF110774.1 AAF14857.1
AF161501.1 AAF29116.1
AF229068.1 AAF82266.1
BC017172.2 AAH17172.1
BC029482.2 AAH29482.1
BC105294.1 AAI05295.1
BE791577.1
CB054126.1 2 312
CN362692.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv951010 Chr.11:63942829 - 64352128 on Build GRCh38 Deletion TRPT1 VEGFB FKBP2 GPR137 MACROD1 PLCB3 ESRRA COX8A CCDC88B OTUB1 TRMT112 NUDT22 TEX40 FERMT3 FLRT1 PRDX5 KCNK4 KCNK4-TEX40 MIR7155 DNAJC4 STIP1 PPP1R14B BAD LOC105369340 NAA40
dgv1963n54 Chr.11:64174262 - 64329761 on Build GRCh38 Loss NUDT22 TRPT1 TEX40 FERMT3 VEGFB FKBP2 GPR137 PRDX5 KCNK4 KCNK4-TEX40 DNAJC4 PLCB3 ESRRA STIP1 PPP1R14B BAD LOC105369340 TRMT112
nsv832186 Chr.11:64204788 - 64402587 on Build GRCh38 Loss NUDT22 TRPT1 TEX40 FERMT3 VEGFB FKBP2 RPS6KA4 GPR137 PRDX5 KCNK4 KCNK4-TEX40 MIR7155 DNAJC4 PLCB3 ESRRA CCDC88B PPP1R14B BAD MIR1237 LOC105369340 TRMT112
nsv469961 Chr.11:64055468 - 64318495 on Build GRCh38 Loss NUDT22 TRPT1 TEX40 FERMT3 FLRT1 VEGFB FKBP2 GPR137 PRDX5 KCNK4 KCNK4-TEX40 DNAJC4 MACROD1 PLCB3 ESRRA STIP1 PPP1R14B BAD LOC105369340 TRMT112

Back To Top

More Information


Set Membership:

Intragenic Exonic Non-exonic DGV Variation

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
regulation of transcription, DNA-templated
transcription initiation from RNA polymerase II promoter
mitochondrion organization
intracellular receptor signaling pathway
response to estradiol
regulation of cell proliferation
steroid hormone mediated signaling pathway
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of transcription from RNA polymerase II promoter
cartilage development
positive regulation of cellular response to insulin stimulus
translational termination
peptidyl-glutamine methylation
tRNA methylation
rRNA methylation
methylation
rRNA (guanine-N7)-methylation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
steroid hormone receptor activity
RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
steroid binding
protein binding
zinc ion binding
protein domain specific binding
sequence-specific DNA binding
protein methyltransferase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline