This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02681836_cn
See other GNB1 CNV Assays ›
Gene Symbol
GNB1
Assay Reference Genome
Location

Chr.1:1785986 on build GRCh38
Cytoband
1p36.33
Assay ID Hs02681836_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

2782

Gene Name:

G protein subunit beta 1

Gene Aliases:

MRD42

Location:

Chr.1:1785285-1891117 on Build GRCh38

Assay Gene Location:

Within Exon 13
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
GNB1 NM_001282538.1 10 2347 NP_001269467.1
NM_001282539.1 11 2444 NP_001269468.1
NM_002074.4 12 2493 NP_002065.1
XM_017001059.1 11 2397 XP_016856548.1
XM_017001060.1 12 2263 XP_016856549.1
XM_017001061.1 11 2436 XP_016856550.1
AF070597.1 4 1462 AAC28652.1
AF070603.1 6 1701 AAC28655.1
AK122949.1 10 2131
BC004186.2 12 2376 AAH04186.1
BC005888.2 10 2144 AAH05888.1
BU684444.1
X04526.1 12 2380 CAA28207.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv5n111 Chr.1:1575421 - 1823403 on Build GRCh38 Deletion MIB2 FNDC10 MMP23B SLC35E2B CDK11A MMP23A CDK11B NADK SLC35E2 GNB1
nsv834091 Chr.1:1701359 - 1880638 on Build GRCh38 Loss CDK11A MMP23A NADK SLC35E2 GNB1
nsv951380 Chr.1:1785162 - 1791261 on Build GRCh38 Deletion GNB1
nsv1160797 Chr.1:1566216 - 1805918 on Build GRCh38 Duplication MIB2 FNDC10 MMP23B SLC35E2B SSU72 CDK11A MMP23A CDK11B NADK SLC35E2 GNB1
nsv482937 Chr.1:10001 - 2368561 on Build GRCh38 Loss LOC107985729 CALML6 MIR6726 GNB1 TNFRSF4 MIR6723 FNDC10 FAM41C SCNN1D SLC35E2B UBE2J2 ATAD3B LOC105378949 CDK11B TTLL10-AS1 FAM138A GABRD FAAP20 LOC105378589 MIR200B TMEM52 MIR6808 ACAP3 MMP23B LOC284600 WASH7P PUSL1 MXRA8 TMEM88B LOC729737 ANKRD65 CPSF3L OR4F16 LOC100287934 CDK11A CCNL2 LOC101928626 MORN1 C1orf159 LINC01128 LINC01342 LOC100130417 MMP23A ATAD3C MIR6859-2 MTND1P23 LOC100129534 LOC107985728 DVL1 AGRN LOC100288175 RNF223 MTND2P28 MRPL20 NOC2L SKI LOC100288069 MIR200A B3GALT6 VWA1 MIR6859-1 AURKAIP1 LOC105378947 MIB2 LOC105378948 MIR6727 TTLL10 LOC100132287 TMEM240 KLHL17 LOC105378591 FAM87B TNFRSF18 LOC100506504 PRKCZ CFAP74 LOC148413 PLEKHN1 LOC102724312 TAS1R3 FAM132A NADK ATAD3A LINC00115 LOC107984841 ISG15 HES4 LOC102725121 LOC100134822 SAMD11 CPTP PERM1 LOC105378592 SLC35E2 OR4F5 OR4F29 MIR1302-2 SSU72 MIR429 DDX11L1 LOC100133331 SDF4
nsv1160926 Chr.1:1772699 - 1905488 on Build GRCh38 Duplication LOC105378949 NADK GNB1
dgv10n100 Chr.1:1779975 - 1884542 on Build GRCh38 Gain NADK GNB1
esv2758913 Chr.1:1389149 - 2092899 on Build GRCh38 Gain+Loss LOC107985729 MRPL20 PRKCZ CFAP74 CALML6 LOC148413 LOC102724312 TMEM88B NADK ATAD3A ANKRD65 GNB1 FNDC10 VWA1 SLC35E2B ATAD3B CDK11A LOC105378949 CCNL2 CDK11B MIB2 MMP23A GABRD ATAD3C LOC105378589 TMEM52 TMEM240 SLC35E2 MMP23B SSU72 LOC105378591

Back To Top

More Information


Set Membership:

Intragenic Exonic DGV Variation

Panther Classification:

Molecular Function -

heterotrimeric G-protein

Gene Ontology Categories:

Function(s) Process(es)

protein folding
signal transduction
adenylate cyclase-activating dopamine receptor signaling pathway
phospholipase C-activating G-protein coupled receptor signaling pathway
positive regulation of cytosolic calcium ion concentration
G-protein coupled acetylcholine receptor signaling pathway
Wnt signaling pathway, calcium modulating pathway
Ras protein signal transduction
cell proliferation
cardiac muscle cell apoptotic process
rhodopsin mediated signaling pathway
platelet activation
sensory perception of taste
retina development in camera-type eye
cellular response to glucagon stimulus
cellular response to prostaglandin E stimulus
cellular response to hypoxia
cellular response to catecholamine stimulus
GTPase activity
signal transducer activity
protein binding
spectrin binding
type 1 angiotensin receptor binding
protein complex binding
GTPase binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-4gvqm:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline