This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02690107_cn
See other CHD1L CNV Assays ›
Gene Symbol
CHD1L
Assay Reference Genome
Location

Chr.1:147187053 on build GRCh38
Cytoband
1q21.1
Assay ID Hs02690107_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

9557

Gene Name:

chromodomain helicase DNA binding protein 1 like

Gene Aliases:

ALC1, CHDL

Location:

Chr.1:147172804-147295766 on Build GRCh38

Assay Gene Location:

Within Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
CHD1L XM_017002858.1 XP_016858347.1

Target Gene Details

Entrez Gene ID:

2330

Gene Name:

flavin containing monooxygenase 5

Gene Aliases:

-

Location:

Chr.1:147183963-147225798 on Build GRCh38

Assay Gene Location:

Within Exon 10
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
FMO5 NM_001144829.2 NP_001138301.1
NM_001144830.2 7 1705 NP_001138302.1
NM_001461.3 9 1838 NP_001452.2
XM_005272946.4 9 1812 XP_005273003.1
XM_005272947.4 9 1556 XP_005273004.1
XM_005272948.4 9 1539 XP_005273005.1
XM_006711245.3 8 1475 XP_006711308.1
XM_011509350.2 9 1843 XP_011507652.1
XM_011509351.2 8 1575 XP_011507653.1
XM_017000801.1 8 1587 XP_016856290.1
XM_017000802.1 8 1357 XP_016856291.1
AK222728.1 9 1553 BAD96448.1
AK225739.1
AK314647.1 9 1766
BC035687.1 7 1378 AAH35687.1
L37080.1 9 1524 AAA67849.1
Z47553.1 9 1530 CAA87633.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
dgv327n100 Chr.1:146028214 - 148860637 on Build GRCh38 Gain MIR5087 CH17-408M7.1 LOC105371242 GPR89B NBPF11 NUDT4P1 RNVU1-7 FMO5 NUDT4P2 MIR6077 LOC101927468 NBPF13P RNVU1-6 LOC100996517 CHD1L GJA5 LOC728989 RNVU1-3 PDE4DIP BCL9 LOC101928979 RNVU1-8 LOC100132057 ACP6 NBPF10 PDZK1P1 LINC01138 NBPF12 RNVU1-1 LOC100505824 RNU1-13P LOC105371226 LINC00624 PDIA3P1 LOC100996763 PPIAL4G HYDIN2 LOC284561 PRKAB2 GJA8 NBPF14 NOTCH2NL
nsv1005055 Chr.1:145642988 - 148991068 on Build GRCh38 Loss MIR5087 RNF115 ANKRD35 CH17-408M7.1 LOC105371242 PDZK1 GPR89B PIAS3 FMO5 NUDT4P2 MIR6077 RNVU1-6 LOC101928979 ACP6 PDZK1P1 NBPF12 RNVU1-1 RNU1-13P LOC105371226 NUDT17 LOC100996763 ANKRD34A RBM8A GNRHR2 LOC284561 PRKAB2 GJA8 POLR3GL MIR6736 GPR89A LIX1L NBPF11 NUDT4P1 RNVU1-7 CD160 LOC101927468 NBPF13P LOC100996517 TXNIP CHD1L GJA5 LOC728989 POLR3C RNVU1-3 PDE4DIP BCL9 RNVU1-8 LOC100132057 NBPF10 LINC01138 LOC100505824 LINC00624 PDIA3P1 PPIAL4G HYDIN2 NBPF14 NOTCH2NL PEX11B HFE2 ITGA10
nsv1121713 Chr.1:147010816 - 147207577 on Build GRCh38 Deletion NBPF13P CHD1L PRKAB2 LOC728989 FMO5 RNVU1-8 PDIA3P1
esv2761299 Chr.1:146068341 - 148926801 on Build GRCh38 Gain+Loss MIR5087 CH17-408M7.1 LOC105371242 GPR89B NBPF11 NUDT4P1 RNVU1-7 FMO5 NUDT4P2 MIR6077 LOC101927468 NBPF13P LOC100996517 CHD1L GJA5 LOC728989 RNVU1-3 PDE4DIP BCL9 RNVU1-8 LOC100132057 ACP6 NBPF10 PDZK1P1 LINC01138 NBPF12 RNVU1-1 LOC100505824 RNU1-13P LOC105371226 LINC00624 PDIA3P1 LOC100996763 PPIAL4G HYDIN2 LOC284561 PRKAB2 GJA8 NBPF14 NOTCH2NL
nsv509457 Chr.1:145948024 - 149021821 on Build GRCh38 Insertion MIR5087 CH17-408M7.1 LOC105371242 LIX1L GPR89B NBPF11 NUDT4P1 RNVU1-7 FMO5 NUDT4P2 MIR6077 LOC101927468 NBPF13P RNVU1-6 LOC100996517 TXNIP CHD1L GJA5 LOC728989 RNVU1-3 PDE4DIP BCL9 LOC101928979 RNVU1-8 LOC100132057 ACP6 NBPF10 PDZK1P1 LINC01138 NBPF12 RNVU1-1 LOC100505824 RNU1-13P LOC105371226 LINC00624 PDIA3P1 LOC100996763 PPIAL4G ANKRD34A HYDIN2 LOC284561 PRKAB2 GJA8 NBPF14 NOTCH2NL HFE2 POLR3GL
nsv1132304 Chr.1:146186094 - 148977674 on Build GRCh38 Duplication MIR5087 CH17-408M7.1 LOC105371242 GPR89B NBPF11 NUDT4P1 RNVU1-7 FMO5 NUDT4P2 MIR6077 LOC101927468 NBPF13P LOC100996517 CHD1L GJA5 LOC728989 RNVU1-3 PDE4DIP BCL9 RNVU1-8 LOC100132057 ACP6 PDZK1P1 LINC01138 NBPF12 RNVU1-1 LOC100505824 RNU1-13P LOC105371226 LINC00624 PDIA3P1 LOC100996763 PPIAL4G HYDIN2 LOC284561 PRKAB2 GJA8 NBPF14 NOTCH2NL
nsv1003688 Chr.1:146036817 - 148934526 on Build GRCh38 Gain MIR5087 CH17-408M7.1 LOC105371242 GPR89B NBPF11 NUDT4P1 RNVU1-7 FMO5 NUDT4P2 MIR6077 LOC101927468 NBPF13P RNVU1-6 LOC100996517 CHD1L GJA5 LOC728989 RNVU1-3 PDE4DIP BCL9 LOC101928979 RNVU1-8 LOC100132057 ACP6 NBPF10 PDZK1P1 LINC01138 NBPF12 RNVU1-1 LOC100505824 RNU1-13P LOC105371226 LINC00624 PDIA3P1 LOC100996763 PPIAL4G HYDIN2 LOC284561 PRKAB2 GJA8 NBPF14 NOTCH2NL
dgv66e199 Chr.1:146055970 - 149056514 on Build GRCh38 Deletion MIR5087 NBPF9 CH17-408M7.1 LOC105371242 GPR89B NBPF11 NUDT4P1 RNVU1-7 FMO5 NUDT4P2 MIR6077 LOC101927468 NBPF13P LOC100996517 CHD1L GJA5 LOC728989 RNVU1-3 PDE4DIP BCL9 LOC101928979 RNVU1-8 LOC100132057 ACP6 NBPF10 PDZK1P1 LINC01138 NBPF12 RNVU1-1 LOC100505824 RNU1-13P LOC105371226 LINC00624 PDIA3P1 LOC100996763 PPIAL4G HYDIN2 LOC284561 PRKAB2 GJA8 NBPF14 NOTCH2NL
nsv437047 Chr.1:147171976 - 147369886 on Build GRCh38 Loss CHD1L PRKAB2 FMO5 PDIA3P1

Back To Top

More Information


Set Membership:

Intragenic Exonic Intronic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

oxidoreductase oxygenase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
biological_process
drug metabolic process
oxidation-reduction process
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity
N,N-dimethylaniline monooxygenase activity
flavin adenine dinucleotide binding
NADP binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline