This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02698595_cn
See other DHDDS CNV Assays ›
Gene Symbol
DHDDS
Assay Reference Genome
Location

Chr.1:26432971 on build GRCh38
Cytoband
1p36.11
Assay ID Hs02698595_cn
Size
Availability Made To Order
Catalog # 4400291
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

79947

Gene Name:

dehydrodolichyl diphosphate synthase subunit

Gene Aliases:

CIT, CPT, DS, HDS, RP59

Location:

Chr.1:26432282-26471306 on Build GRCh38

Assay Gene Location:

Overlaps Exon 2 - Intron 2
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
DHDDS NM_001243564.1 NP_001230493.1
NM_001243565.1 NP_001230494.1
NM_001319959.1 NP_001306888.1
NM_024887.3 NP_079163.2
NM_205861.2 NP_995583.1
XM_006710912.2 XP_006710975.1
XM_006710913.2 XP_006710976.1
XM_006710914.2 XP_006710977.1
XM_006710915.2 XP_006710978.1
XM_006710916.2 XP_006710979.1
XM_006710917.2 XP_006710980.1
XM_006710918.2 XP_006710981.1
XM_011542183.2 XP_011540485.1
XM_011542184.2 XP_011540486.1
XM_011542185.2 XP_011540487.1
XM_011542186.2 XP_011540488.1
XM_017002379.1 XP_016857868.1
XM_017002380.1 XP_016857869.1
XM_017002381.1 XP_016857870.1
XM_017002382.1 XP_016857871.1
XM_017002383.1 XP_016857872.1
XM_017002384.1 XP_016857873.1
AB090852.1 BAC57588.1
AK023164.1 BAB14439.1
AK297134.1
AK308187.1
AK316485.1
BC003643.2 AAH03643.1
BC004117.2 AAH04117.1
BX648507.1
DA756178.1
GU727641.1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic non-DGV Variation

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein glycosylation
dolichyl diphosphate biosynthetic process
polyprenyltransferase activity
protein binding

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0