This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs02744370_cn
See other KIFC3 CNV Assays ›
Gene Symbol
KIFC3
Assay Reference Genome
Location

Chr.16:57772256 on build GRCh38
Cytoband
16q21
Assay ID Hs02744370_cn
Size
Availability Made To Order
Catalog # 4400291
Price 4,496.00
Your Price
Online offer:
4496.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Target Gene Details

Entrez Gene ID:

3801

Gene Name:

kinesin family member C3

Gene Aliases:

-

Location:

Chr.16:57758217-57863053 on Build GRCh38

Assay Gene Location:

Overlaps Intron 16 - Exon 17
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
KIFC3 NM_001130099.1 NP_001123571.1
NM_001130100.1 NP_001123572.1
NM_001318710.1 NP_001305639.1
NM_001318711.1 NP_001305640.1
NM_001318712.1 NP_001305641.1
NM_001318713.1 NP_001305642.1
NM_001318714.1 NP_001305643.1
NM_001318715.1 NP_001305644.1
NM_005550.3 NP_005541.3
NR_134678.1
XM_005255937.1 XP_005255994.1
XM_005255944.1 XP_005256001.1
XM_006721188.1 XP_006721251.1
XM_011523075.1 XP_011521377.1
XM_011523076.1 XP_011521378.1
XM_011523077.1 XP_011521379.1
XM_011523078.1 XP_011521380.1
XM_011523079.1 XP_011521381.1
XM_017023219.1 XP_016878708.1
XM_017023220.1 XP_016878709.1
XM_017023221.1 XP_016878710.1
XM_017023222.1 XP_016878711.1
XM_017023223.1 XP_016878712.1
XM_017023224.1 XP_016878713.1
XM_017023225.1 XP_016878714.1
XM_017023226.1 XP_016878715.1
XM_017023227.1 XP_016878716.1
AB209290.1 BAD92527.1
AK291737.1
AK295926.1
AK296995.1
AK299431.1
AK302743.1
AK303021.1
AK304132.1
AK307846.1
BC001211.1
BC034234.1 AAH34234.1
BC041132.1 AAH41132.1
DC381406.1
DQ426879.1
DQ426880.1

Target Gene Details

Entrez Gene ID:

102465463

Gene Name:

microRNA 6772

Gene Aliases:

hsa-mir-6772

Location:

Chr.16:57772289-57772352 on Build GRCh38

Assay Gene Location:

Overlaps - Exon 1
Gene Symbol Transcript Accession Exon Location Assay Transcript Location Protein ID
MIR6772 NR_106830.1

Target Copy Number Variation Details

DGV Version:

Release date: 2016-05-15, GRCh GRCh38
Target
Variation
Location CNV
Subtype
Genes
nsv523237 Chr.16:57753115 - 57786197 on Build GRCh38 Loss MIR6772 KIFC3 KATNB1
nsv471089 Chr.16:57722731 - 57788355 on Build GRCh38 Loss MIR6772 KIFC3 KATNB1 DRC7
nsv525464 Chr.16:57714251 - 57832787 on Build GRCh38 Gain LOC388282 MIR6772 KIFC3 KATNB1 DRC7
nsv509622 Chr.16:57720104 - 57826484 on Build GRCh38 Insertion LOC388282 MIR6772 KIFC3 KATNB1 DRC7
nsv528250 Chr.16:57715692 - 57786197 on Build GRCh38 Loss MIR6772 KIFC3 KATNB1 DRC7
nsv952038 Chr.16:57734189 - 57774488 on Build GRCh38 Deletion MIR6772 KIFC3 KATNB1

Back To Top

More Information


Set Membership:

Intragenic Non-exonic DGV Variation

Panther Classification:

Molecular Function -

microtubule binding motor protein

Gene Ontology Categories:

Function(s) Process(es)

microtubule-based movement
Golgi organization
visual perception
zonula adherens maintenance
epithelial cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, minus-end-directed
ATPase activity

Back To Top

Related Products

  • TaqMan® Genotyping Master Mix
  • TaqPath®ProAmp® Master Mix
  • TaqMan® Universal Master Mix II, no UNG
  • TaqMan® Gene Expression Master Mix
  • TaqMan® Universal PCR Master Mix
  • TaqMan® Universal PCR Master Mix, No AmpErase UNG

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-nlplw:80/100.66.79.197:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline